Skip to Content
Merck
All Photos(1)

Key Documents

294926

Sigma-Aldrich

1-(1-Naphthyl)ethylamine

98%

Synonym(s):

(±)-1-(1-Naphthyl)ethylamine, (±)-α-Methyl-1-naphthalenemethylamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C10H7CH(CH3)NH2
CAS Number:
Molecular Weight:
171.24
Beilstein:
3197375
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

refractive index

n20/D 1.621 (lit.)

bp

156 °C/15 mmHg (lit.)

density

1.063 g/mL at 25 °C (lit.)

SMILES string

CC(N)c1cccc2ccccc12

InChI

1S/C12H13N/c1-9(13)11-8-4-6-10-5-2-3-7-12(10)11/h2-9H,13H2,1H3

InChI key

RTCUCQWIICFPOD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

The adsorption of 1-(1-naphthyl)ethylamine on platinum surfaces has been characterized by reflection-absorption infrared spectroscopy and temperature-programmed desorption both under ultrahigh vacuum and in situ from liquid solutions.

Application

(R)1-(1-Naphthyl)ethylamine, a new chiral modifier, has been employed in the enantioselective hydrogenation of ethyl pyruvate to ethyl lactate over 5 wt% Pt/Al2O3.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Enantioselective Hydrogenation of Ethyl Pyruvate over Pt/Alumina Modified by (R)-1-(1-Naphthyl) ethylamine Derivatives.
Minder B, et al.
J. Catal., 160(2), 261-268 (1992)
Ilkeun Lee et al.
Journal of the American Chemical Society, 130(44), 14597-14604 (2008-10-14)
The adsorption of 1-(1-naphthyl)ethylamine (NEA) on platinum surfaces has been characterized by reflection-absorption infrared spectroscopy (RAIRS) and temperature-programmed desorption (TPD) both under ultrahigh vacuum and in situ from liquid solutions. The main focus of this study was to identify the
Jian Tang et al.
Analytica chimica acta, 946, 96-103 (2016-11-09)
In this work, a detail study has been performed on the enantioselectivity of per(3-chloro-4-methyl)phenylcarbamate-β-CD clicked chiral stationary phase (CSP) in high-performance liquid chromatography. Both normal phase and polar organic mobile phases have been explored for the enantioseparation of 39 model
Akash Singh et al.
Advanced materials (Deerfield Beach, Fla.), 33(3), e2005868-e2005868 (2020-12-08)
Crystalline metal halide perovskites (MHPs) have provided unprecedented advances in interdisciplinary fields of materials, electronics, and photonics. While crystallinity offers numerous advantages, the ability to access a glassy state with distinct properties provides unique opportunities to extend the associated structure-property
A Ishii et al.
Science advances, 6(46) (2020-11-13)
Detection of circularly polarized light (CPL) has a high potential for development of various optical technologies. Conventional photodetectors require optical polarizers on the device to detect polarized light, and this causes substantial losses of sensitivity and resolution in light detection.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service