Skip to Content
Merck
All Photos(2)

Key Documents

293296

Sigma-Aldrich

Butyl benzoate

99%

Synonym(s):

Benzoic acid n-butyl ester, n-Butyl benzoate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5COOCH2CH2CH2CH3
CAS Number:
Molecular Weight:
178.23
Beilstein:
1867073
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Assay

99%

refractive index

n20/D 1.498 (lit.)

bp

249 °C (lit.)

density

1.01 g/mL at 25 °C (lit.)

functional group

ester
phenyl

SMILES string

CCCCOC(=O)c1ccccc1

InChI

1S/C11H14O2/c1-2-3-9-13-11(12)10-7-5-4-6-8-10/h4-8H,2-3,9H2,1H3

InChI key

XSIFPSYPOVKYCO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

Butyl benzoate has been used as:
  • preservative in commercial cosmetic lotions[1]
  • as involatile solvent, for sampling of isocyanates with silica gel solid phase extraction[2]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

222.8 °F - closed cup

Flash Point(C)

106 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

E Marengo et al.
Journal of chromatographic science, 39(8), 339-344 (2001-08-22)
An ion-interaction high-performance liquid chromatography (HPLC)-diode-array detection method is developed and optimized for the separation of typical antimicrobial agents used in cosmetics and hygiene products. The most used preservatives contain different molecular structures, different functionalities, and are characterized by different
John White
The Annals of occupational hygiene, 50(1), 15-27 (2005-08-30)
Isocyanates (NCO) are potent sensitizers and the largest cause of occupational asthma in the UK. Probably, MDHS 25/3 is the method most commonly used worldwide for the determination of organic NCO in air. This method uses an electrochemical (EC) detector
Philipp Denk et al.
Analytical and bioanalytical chemistry, 410(14), 3395-3404 (2018-04-07)
Elastic therapeutic tapes are an important tool in the field of physical therapy and medicine. These tapes contain types of adhesive. However, sensory evaluations revealed the release of pronounced and irritating odors of the tapes. Negative odors were, amongst others
Suvi-Katriina Ruokonen et al.
Journal of chromatography. A, 1405, 178-187 (2015-06-15)
The effect of three phosphonium and imidazolium ionic liquids (ILs) on the interaction between liposomes and common pharmaceuticals found in wastewaters was studied. The liposomes comprised zwitterionic phosphatidyl choline and negatively charged phosphatidyl glycerol. A set of common cationic, anionic

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service