190195
1-Ethylpropylamine
97%
Synonym(s):
3-Aminopentane
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
(C2H5)2CHNH2
CAS Number:
Molecular Weight:
87.16
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Quality Level
Assay
97%
refractive index
n20/D 1.4055 (lit.)
bp
89-91 °C (lit.)
density
0.748 g/mL at 25 °C (lit.)
functional group
amine
SMILES string
CCC(N)CC
InChI
1S/C5H13N/c1-3-5(6)4-2/h5H,3-4,6H2,1-2H3
InChI key
PQPFFKCJENSZKL-UHFFFAOYSA-N
Application
1-Ethylpropylamine has been used:
- in the synthesis of (1-ethyl-propyl)-[3-(4-methoxy-2-methylphenyl)-2-methyl-6,7-dihydro-5H-1,4,8a-triaza-s-indacen-8-yl]-amine
- in the synthesis of monoclinic VPO4·H2O
- as internal standard in the screening of aziridine and 2-chloroethylamine in active pharmaceutical ingredients by gas chromatography-mass spectrometry.
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Flam. Liq. 2 - Skin Corr. 1B
Storage Class Code
3 - Flammable liquids
WGK
WGK 3
Flash Point(F)
35.6 °F - closed cup
Flash Point(C)
2 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Reaction of vanadium phosphates with alcohols at elevated temperature and pressure.
Dong W-S, et al.
Journal of Materials Chemistry, 15(31), 3214-3220 (2005)
Julián Zapata et al.
Journal of pharmaceutical and biomedical analysis, 55(3), 458-465 (2011-03-23)
A simple, fast and fully automated method for the screening of aziridine (AZD) and 2-chloroethylamine (CEA) in active pharmaceutical ingredients (API) has been developed. The method is based on the in-fiber derivatization of the amines extracted from the sample headspace
Tetsuji Saito et al.
Bioorganic & medicinal chemistry, 19(18), 5432-5445 (2011-08-26)
To identify an orally active corticotropin-releasing factor 1 receptor antagonist, a series of 6,7-dihydro-5H-cyclopenta[d]pyrazolo[1,5-a]pyrimidines and their derivatives were designed, synthesized and evaluated. An in vitro study followed by in vivo and pharmacokinetic studies of these heterotricyclic compounds led us to
Jan Leipert et al.
Lab on a chip, 19(20), 3490-3498 (2019-09-19)
While LC-MS-based proteomics with high nanograms to micrograms of total protein has become routine, the analysis of samples derived from low cell numbers is challenged by factors such as sample losses, or difficulties encountered with the manual manipulation of small
Jan Leipert et al.
Analytical and bioanalytical chemistry, 410(19), 4737-4748 (2018-02-23)
The identification and quantification of molecules involved in bacterial communication are major prerequisites for the understanding of interspecies interactions at the molecular level. We developed a procedure allowing the determination of 2-heptyl-4(1H)-quinolone (HHQ) and 2-heptyl-3-hydroxy-4(1H)-quinolone (PQS) and the virulence factor
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service