Skip to Content
Merck
All Photos(2)

Key Documents

184462

Sigma-Aldrich

Butyl chloroformate

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClCOOCH2CH2CH2CH3
CAS Number:
Molecular Weight:
136.58
Beilstein:
605635
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
form:
liquid
Assay:
98%

vapor pressure

2.42 psi ( 20 °C)

Quality Level

Assay

98%

form

liquid

refractive index

n20/D 1.412 (lit.)

bp

142 °C (lit.)

density

1.074 g/mL at 25 °C (lit.)

functional group

chloro

storage temp.

2-8°C

SMILES string

CCCCOC(Cl)=O

InChI

1S/C5H9ClO2/c1-2-3-4-8-5(6)7/h2-4H2,1H3

InChI key

NRDQFWXVTPZZAZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

  • Butyl chloroformate was used in the synthesis of dextran alkyl carbonates.
  • It was also used in the synthesis of S-tert-alkyl-N,N-alkoxycarbonylmethyl-dithiocarbamate, radical polymerization with reversible addition-fragmentation chain transfer agent.
  • It was used as derivatization reagent in determination of linsidomine in plasma using high-performance liquid chromatographic separation with tandem mass spectrometric detection.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Flam. Liq. 3 - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

100.4 °F - closed cup

Flash Point(C)

38 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Multiblock copolymer synthesis via controlled radical polymerization in aqueous dispersions. Part 1: Synthesis of S-tert-alkyl-N,N-alkoxycarbonylmethyl-dithiocarbamate.
Bussels R and Koning CE.
Tetrahedron, 61(5), 1167-1174 (2005)
F C Sutherland et al.
Journal of pharmaceutical and biomedical analysis, 22(3), 461-467 (2000-04-15)
A sensitive method for the determination of linsidomine in plasma was developed, using high-performance liquid chromatographic (HPLC) separation with tandem mass spectrometric detection. Linsidomine was derivatised with propyl chloroformate and extracted with tert-butyl methyl ether/1,2-dichloroethane (55:45, v/v), back-extracted into HCl
Andrea Y Garzón-Serrano et al.
Journal of forensic sciences, 65(4), 1085-1093 (2020-03-17)
Ammonium nitrate fuel oil is an explosive mixture found in most antipersonnel landmines (APL) buried throughout the Colombian territory. During more than 50 years of internal conflict, the Colombian government has found that trained dogs are the most effective method to
Tomáš Hložek et al.
Journal of analytical toxicology, 40(9), 749-753 (2016-09-04)
A simple, sensitive and robust method for simultaneous determination of antiepileptic drugs (gabapentin, pregabalin and vigabatrin) in human serum using GC-MS was developed and validated for clinical toxicology purposes. This method employs an emerging class of derivatization agents - alkyl
Thomas Elschner et al.
Carbohydrate polymers, 93(1), 216-223 (2013-03-08)
Dextran alkyl carbonates were synthesized applying ethyl chloroformate, butyl chloroformate, butyl fluoroformate and 1H-imidazole-1-carboxylates. The influence of the reaction conditions on the reaction efficiency and the substitution pattern was studied in detail. The structure of the products obtained was clearly

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service