Skip to Content
Merck
All Photos(2)

Key Documents

182338

Sigma-Aldrich

Poly(octadecyl methacrylate) solution

average Mw ~170,000 by GPC, in toluene

Synonym(s):

Poly(octadecyl methacrylate), Poly(stearyl methacrylate)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C22H42O2)n
CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23
Pricing and availability is not currently available.

form

liquid

Quality Level

mol wt

average Mw ~170,000 by GPC

concentration

in toluene
30-35 wt. % (in toluene)

weight

Unit weight does not include weight of solvent.

transition temp

Tg −100 °C
Tm 37.5 °C

SMILES string

CCCCCCCCCCCCCCCCCCOC(=O)C(C)=C

InChI

1S/C22H42O2/c1-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20-24-22(23)21(2)3/h2,4-20H2,1,3H3

InChI key

HMZGPNHSPWNGEP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Poly(octadecyl methacrylate) may be used to prepare block copolymers to form molecular brushes.[1][2]

Other Notes

Unit weight does not include weight of solvent.

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 3 - Asp. Tox. 1 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

39.2 °F - closed cup

Flash Point(C)

4.0 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis and visualization of densely grafted molecular brushes with crystallizable poly (octadecyl methacrylate) block segments.
Qin S, et al.
Macromolecules, 36(3), 605-612 (2003)
Orientation of poly (octadecyl methacrylate) and poly (octadecyl acrylate) in Langmuir-Blodgett monolayers investigated by polarized infrared spectroscopy.
Mumby S J, et al.
Macromolecules, 19(4), 1054-1059 (1986)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service