Merck
Chitin dipentanoate as the new technologically usable biomaterial.
Materials science & engineering. C, Materials for biological applications (2015-06-29)
Karolina Skołucka-Szary, Aleksandra Ramięga, Wanda Piaskowska, Bartosz Janicki, Magdalena Grala, Piotr Rieske, Ewelina Stoczyńska-Fidelus, Sylwester Piaskowski
ABSTRACT
In this article, the synthesis of novel biopolymer, chitin dipentanoate (Di-O-Valeryl Chitin, DVCH) has been described. DVCH is a chitin derivative esterified with two valeryl groups at positions 3 and 6 of the N-acetylglucosamine units and it is soluble in common organic solvents like ethanol, methanol, acetone, dichloromethane, 1,2-dichloroethane, N,N-dimethylmethanamide, N,N-dimethylacetamide and ethyl acetate. Highly efficient synthesis (degree of esterification close to 2) of DVCH was achieved by employing a huge excess of valeric anhydride used as both the acylation agent and the reaction medium in the presence of perchloric acid as catalyst. Studies on the DVCH synthesis were aimed at finding optimal conditions (temperature, reaction time) to obtain DVCH with high reaction yield and desirable physicochemical properties. Biological data demonstrate that DVCH is non-cytotoxic in vitro and doesn't exert irritating or allergic effects to animal skin. Thanks to its filmogenic properties, it can be used to manufacture threads, foils, foams and non-woven materials. Moreover, DVCH can be easily processed by salt-leaching method to prepare highly porous structures exhibiting open-cell architecture, that can be further employed in wound dressing therapies and scaffolds for tissue engineering.
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
Acetone-d6, 99.9 atom % D
Sigma-Aldrich
Acetone-d6, "100%", 99.95 atom % D
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
HEPES, BioUltra, for molecular biology, ≥99.5% (T)
Sigma-Aldrich
Acetone-d6, 99.9 atom % D, contains 0.03 % (v/v) TMS
Sigma-Aldrich
Acetone-d6, 99.5 atom % D
Sigma-Aldrich
Acetone-d6, 99.9 atom % D, contains 1 % (v/v) TMS
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
Sodium bicarbonate, BioXtra, 99.5-100.5%
Sigma-Aldrich
Sodium bicarbonate, powder, BioReagent, for molecular biology, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
SAFC
HEPES
SAFC
HEPES
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Acetic acid, for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
HEPES, anhydrous, free-flowing, Redi-Dri, ≥99.5%