Skip to Content
Merck
All Photos(4)

Key Documents

L5750

Sigma-Aldrich

Sodium dodecyl sulfate

92.5-100.5% based on total alkyl sulfate content basis

Synonym(s):

Dodecyl sodium sulfate, Dodecyl sulfate sodium salt, Lauryl sulfate sodium salt, SDS, Sodium lauryl sulfate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)11OSO3Na
CAS Number:
Molecular Weight:
288.38
Beilstein:
3599286
EC Number:
MDL number:
UNSPSC Code:
12161900
eCl@ss:
39093306
PubChem Substance ID:
NACRES:
NA.21

description

anionic

Quality Level

Assay

92.5-100.5% based on total alkyl sulfate content basis

mol wt

288.38 g/mol

aggregation number

62

CMC

7-10 mM (20-25°C)

mp

204-207 °C (lit.)

transition temp

cloud point >100 °C

solubility

water: soluble

HLB

40

SMILES string

[Na+].CCCCCCCCCCCCOS([O-])(=O)=O

InChI

1S/C12H26O4S.Na/c1-2-3-4-5-6-7-8-9-10-11-12-16-17(13,14)15;/h2-12H2,1H3,(H,13,14,15);/q;+1/p-1

InChI key

DBMJMQXJHONAFJ-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Anionic detergent
Sodium dodecyl sulfate has been used:
  • to dissolve insoluble purple formazan in 3-(4,5-dimethylthiazol-2-yl)-2,5-diphenyl tetrazolium bromide (MTT) assay
  • to perform SDS–PAGE of yeast tubulin
  • as an additive in linolenic acid solution to assess the total antioxidative activity (TAA) of the bacteria using the linolenic acid test (LA-test)

Biochem/physiol Actions

Sodium dodecyl sulfate (SDS)/lauryl sulfate, an ionic detergent has the ability to distract biological membranes. It can block RNase and deoxyribonuclease (DNase) activity. SDS is one of the main constituents of several reagents, that are used to purify nucleic acids. It is also considered as a permeability enhancer in oral formulations.

Quality

Predominantly (approx. 70%) C12; lauryl sulfate, sodium salt; balance consists of higher homologs.

Analysis Note

Actual composition given on label

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

338.0 °F

Flash Point(C)

170 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The two alpha-tubulin isotypes in budding yeast have opposing effects on microtubule dynamics in vitro
Bode CJ, et al.
EMBO Reports, 4(1), 94-99 (2003)
Resilient Ribonucleases
RNA Methodologies (2010)
Two antioxidative lactobacilli strains as promising probiotics
Kullisaar T, et al.
International Journal of Food Microbiology, 72(3), 215-224 (2002)
Safety and efficacy of self-assembling bubble carriers stabilized with sodium dodecyl sulfate for oral delivery of therapeutic proteins
Lin P Y, et al.
Journal of Controlled Release : Official Journal of the Controlled Release Society, 259, 168-175 (2017)
Ursolic, oleanolic and betulinic acids: antibacterial spectra and selectivity indexes.
Fontanay S, et al.
Journal of Ethnopharmacology, 120(2), 272-276 (2008)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service