Merck
  • Differences between Solution and Membrane Forms of Chitosan on the In Vitro Activity of Fibroblasts.
Differences between Solution and Membrane Forms of Chitosan on the In Vitro Activity of Fibroblasts.
Balkan medical journal (2015-03-12)
Bahar Uslu, Burcu Biltekin, Seçnur Denir, Suna Özbaş-Turan, Serap Arbak, Jülide Akbuğa, Ayhan Bilir
ABSTRACT
Chitosan, a linear polysaccharide, has been recently used in biomedical applications. In vitro studies have demonstrated its effect on cellular growth and its stimulatory action on cellular layer formation. The present study aims to compare the proliferative effects of chitosan in two forms, membranous and solution forms, on Swiss 3T3 mouse embryonic fibroblasts. In vitro study. Three experimental groups were formed: cells were cultured in a normal medium without chitosan (Control Group); cells were cultured either in a medium containing 2.0% chitosan in membranous form (Membrane Group) or chitosan solution at a concentration of 2.0% (Solution Group). Two different methods were used in the experiments: cells cultured on the medium containing chitosan in solution or membranous forms (method 1); and chitosan solution or membranous forms were added into the medium containing previously cultured cells (method 2). Scanning electron microscopic investigations of the experimental groups revealed cells with well-defined cellular projections, intact cellular membranes and tight intercellular junctions. They were especially prominent in the membrane group of method 1 and in the membrane and solution groups of method 2. Mouse monoclonal anti-collagen 1 primary antibody was used to indicate collagen synthesis. Prominent collagen synthesis was detected in the membrane groups on the 10(th) day of culture for both methods. Bromodeoxyuridine (BrdU) and MTT assays were performed in order to assess cellular proliferation and viability, respectively. BrdU labelling tests indicated a higher proliferation index in the membrane group of method 1 on the 5(th) and 10(th) days. For the second method, the membranous form on the 10(th) day and solution form on the 5(th) day were the most effective groups in terms of cellular proliferation. MTT results reflected a high cellular viability in method 1 on the 5(th) day of treatment with the membranous form, whereas cellular viability was highest in the solution form of method 2 on the 5(th) day. The membranous form of chitosan induced a significant proliferative effect and increased the ratio of cell-to-cell junctions of Swiss 3T3 mouse embryonic fibroblasts. Conveniently, the solution form also resulted in enhanced cell proliferation and viability compared to the control group. As the solution form is easy to prepare and apply to cells compared to the membrane form, the application of Chitosan directly to media appears to be a convenient alternative for tissue engineering approaches.
Sigma-Aldrich
Glutaraldehyde solution, technical, ~50% in H2O (5.6 M)
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Sigma-Aldrich
Glutaraldehyde solution, 50 wt. % in H2O
Sigma-Aldrich
Pentyl acetate, puriss. p.a., ≥98.5% (GC)
Sigma-Aldrich
Pentyl acetate, 99%
Sigma-Aldrich
Glutaric dialdehyde solution, 50 wt. % in H2O, FCC
Sigma-Aldrich
Amyl acetate, ≥99%, FG
Sigma-Aldrich
Osmium tetroxide, Sealed ampule.
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)
Sigma-Aldrich
Glutaraldehyde solution, Grade II, 25% in H2O
Sigma-Aldrich
Triton X-305 solution, 70% in H2O
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 70% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, 98%
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 25% in H2O, specially purified for use as an electron microscopy fixative
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 50% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Os EnCat® 40, extent of labeling: 0.3 mmol/g Os loading
Supelco
Dehydrated Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 8% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Glutaraldehyde solution, 50% in H2O, suitable for photographic applications
USP
Dehydrated Alcohol, United States Pharmacopeia (USP) Reference Standard
Supelco
Pentyl acetate, analytical standard
Sigma-Aldrich
Osmium tetroxide, ReagentPlus®, 99.8%
Sigma-Aldrich
Osmium tetroxide, ACS reagent, ≥98.0%
Supelco
Dimethyl sulfoxide, for inorganic trace analysis, ≥99.99995% (metals basis)
Sigma-Aldrich
Ethanol, tested according to Ph. Eur.
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Osmium tetroxide solution, suitable for electron microscopy, 4% in H2O
Sigma-Aldrich
Osmium tetroxide solution, suitable for electron microscopy, 2% in H2O
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture