Skip to Content
Merck
All Photos(1)

Key Documents

66962

Supelco

Pentyl acetate

analytical standard

Synonym(s):

Amyl acetate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3COO(CH2)4CH3
CAS Number:
Molecular Weight:
130.18
Beilstein:
1744753
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

vapor density

4.5 (vs air)

vapor pressure

4 mmHg ( 20 °C)

Assay

≥97.0% (GC)

autoignition temp.

680 °F

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.402 (lit.)
n20/D 1.403

bp

142-149 °C (lit.)

mp

−100 °C (lit.)

density

0.876 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

neat

SMILES string

CCCCCOC(C)=O

InChI

1S/C7H14O2/c1-3-4-5-6-9-7(2)8/h3-6H2,1-2H3

InChI key

PGMYKACGEOXYJE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
Banana essence. Used as test odorant in studies of olfactory function and in studies of the psychosocial effects of odor.

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

105.8 °F

Flash Point(C)

41 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Guoxin Ying et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 34(18), 6377-6388 (2014-05-03)
Centrins are ancient calmodulin-related Ca(2+)-binding proteins associated with basal bodies. In lower eukaryotes, Centrin2 (CETN2) is required for basal body replication and positioning, although its function in mammals is undefined. We generated a germline CETN2 knock-out (KO) mouse presenting with
Elizabeth Steuer et al.
Journal of neurotrauma, 31(14), 1277-1291 (2014-04-04)
Loss of olfactory function is an early indicator of traumatic brain injury (TBI). The regenerative capacity and well-defined neural maps of the mammalian olfactory system enable investigations into the degeneration and recovery of neural circuits after injury. Here, we introduce
Emanuele Brai et al.
The European journal of neuroscience, 40(10), 3436-3449 (2014-09-23)
Notch signalling plays an important role in synaptic plasticity, learning and memory functions in both Drosophila and rodents. In this paper, we report that this feature is not restricted to hippocampal networks but also involves the olfactory bulb (OB). Odour
Matteo Gianella-Borradori et al.
Bioorganic & medicinal chemistry, 23(1), 241-263 (2014-12-10)
The cannabinoid receptor 2 (CB2R) has been linked with the regulation of inflammation, and selective receptor activation has been proposed as a target for the treatment of a range of inflammatory diseases such as atherosclerosis and arthritis. In order to
Bahar Uslu et al.
Balkan medical journal, 32(1), 69-78 (2015-03-12)
Chitosan, a linear polysaccharide, has been recently used in biomedical applications. In vitro studies have demonstrated its effect on cellular growth and its stimulatory action on cellular layer formation. The present study aims to compare the proliferative effects of chitosan

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service