Merck
Naphthalenyl derivatives for hitting P-gp/MRP1/BCRP transporters.
Bioorganic & medicinal chemistry (2013-01-26)
Nicola A Colabufo, Marialessandra Contino, Mariangela Cantore, Elena Capparelli, Maria Grazia Perrone, Giuseppe Cassano, Giuseppe Gasparre, Marcello Leopoldo, Francesco Berardi, Roberto Perrone
ABSTRACT
Substituted naphthalenyl derivatives bearing oxazole, or thiazole or furyl heteronuclei have been carried out as bioisosters of aryl-oxazoles and -thiazoles derivatives previously reported in order to investigate the role of the hindrance on the activity towards P-gp/BCRP/and MRP1 transporters. In addition, the role of naphthalenyl group to modulate P-gp intrinsic activity of these compounds was ascertained. The results demonstrated that all naphthalenyl derivatives displayed comparable P-gp activity with respect to lead compounds previously characterized in our SAR studies but were less active towards BCRP and MRP1 pumps. In terms of intrinsic activity, the replacement of aryl with naphthalenyl moiety led to P-gp inhibitors, unambiguous or ambiguous substrates on the base of the heteronucleus and the substituent on the naphthalenyl fragment. Indeed, oxazole derivatives were: inhibitors (R=H, F, OH), unambiguous substrates (R=OCH(3)), or ambiguous substrate (R=Br); thiazole derivatives were: unambiguous substrates (R=OCH(3), Br), or ambiguous substrates (R=H, F). Finally furyl derivatives were ambiguous substrates.
Product Description
Supelco
Naphthalene solution, certified reference material, 5000 μg/mL in methanol
Supelco
Naphthalene, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Naphthalene, analytical standard
Sigma-Aldrich
Naphthalene, 99%
Sigma-Aldrich
Naphthalene, suitable for scintillation, ≥99%
Supelco
Melting point standard 79-81°C, analytical standard
Sigma-Aldrich
Rhodamine 123, mitochondrial specific fluorescent dye
Sigma-Aldrich
Rhodamine 123, BioReagent, for fluorescence, ≥85% (HPLC)