Merck
  • Recombinant hepatocyte growth factor treatment in a canine model of congenital liver hypoplasia.
Recombinant hepatocyte growth factor treatment in a canine model of congenital liver hypoplasia.
Liver international : official journal of the International Association for the Study of the Liver (2011-07-08)
Hedwig S Kruitwagen, Brigitte Arends, Bart Spee, Bas Brinkhof, Ted S G A M van den Ingh, Victor P M G Rutten, Louis C Penning, Tania Roskams, Jan Rothuizen
ABSTRACT
Although the liver has a large regenerative capacity, in many hepatopathies, these repair mechanisms fail. The therapeutic potential of hepatocyte growth factor (HGF) has been proven in numerous toxin-induced liver failure models in rodents, but never in spontaneously occurring liver diseases in larger animal models. The aim of this study was to induce liver growth in a hypoplastic liver by the administration of exogenous recombinant HGF. The natural hypoplastic liver model used is the canine congenital portosystemic shunt (CPSS) characterized by strongly reduced liver growth and function. Recombinant HGF (rHGF), 200 μg/kg, was given twice daily during 3 weeks by an intravenous injection in six dogs with CPSS. Liver volumes were determined by computed tomography before and at 1, 2, 3 and 7 weeks after the initiation of treatment. Portosystemic shunting was evaluated with an ammonia tolerance test and liver portal perfusion was quantified with scintigraphy. Simultaneously, blood parameters for liver function were assayed and liver biopsies were taken for histology, immunohistochemistry and gene-expression measurements. During 3 weeks of HGF treatment, hepatocyte proliferation increased and an increase in liver volume up to 44% was seen, persisting in two dogs up to 4 weeks after the termination of treatment. Ki-67 expression, gene expression of E2F1 and CDC6, phosphorylated-c-MET and phosphorylated-ERK1/2 protein levels confirmed increased hepatocyte proliferation and HGF signalling. The aberrant portal perfusion did not change during treatment. Transient in vivo liver growth is shown using CPPS as a naturally occurring large animal model, indicating the therapeutic potential of HGF in liver disease.
Sigma-Aldrich
Anti-Hepatocyte Growth Factor Receptor (c-Met) antibody produced in goat, affinity isolated antibody, lyophilized powder