Merck
  • Fms-like tyrosine kinase 3 is a regulator of the cardiac side population in mice.
Fms-like tyrosine kinase 3 is a regulator of the cardiac side population in mice.
Life science alliance (2021-12-15)
Giacomo Della Verde, Michika Mochizuki, Vera Lorenz, Julien Roux, Lifen Xu, Leandra Ramin-Wright, Otmar Pfister, Gabriela M Kuster
ABSTRACT
Fms-like tyrosine kinase 3 (Flt3) is a regulator of hematopoietic progenitor cells and a target of tyrosine kinase inhibitors. Flt3-targeting tyrosine kinase inhibitors can have cardiovascular side effects. Flt3 and its ligand (Flt3L) are expressed in the heart, but little is known about their physiological functions. Here, we show that cardiac side population progenitor cells (SP-CPCs) from mice produce and are responsive to Flt3L. Compared with wild-type, flt3L-/- mice have less SP-CPCs with less contribution of CD45-CD34+ cells and lower expression of genes related to epithelial-to-mesenchymal transition, cardiovascular development and stem cell differentiation. Upon culturing, flt3L-/- SP-CPCs show increased proliferation and less vasculogenic commitment, whereas Akt phosphorylation is lower. Notably, proliferation and differentiation can be partially restored towards wild-type levels in the presence of alternative receptor tyrosine kinase-activating growth factors signaling through Akt. The lower vasculogenic potential of flt3L-/- SP-CPCs reflects in decreased microvascularisation and lower systolic function of flt3L-/- hearts. Thus, Flt3 regulates phenotype and function of murine SP-CPCs and contributes to cellular and molecular properties that are relevant for their cardiovasculogenic potential.
Sigma-Aldrich
Bovine Serum Albumin, heat shock fraction, pH 7, ≥98%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Akt Inhibitor VIII, Isozyme-Selective, Akti-1/2, Akt Inhibitor VIII, Isozyme-Selective, Akti-1/2, CAS 612847-09-3, is a cell-permeable, reversible inhibitor of Akt1/Akt2 (IC₅₀ = 58 nM, 210 nM, & 2.12 µM for Akt1, Akt2, and Akt3, respectively).