Merck
  • Response and resistance to CDK12 inhibition in aggressive B-cell lymphomas.
Response and resistance to CDK12 inhibition in aggressive B-cell lymphomas.
Haematologica (2021-06-25)
Jing Gao, Michelle Y Wang, Yuan Ren, Tint Lwin, Tao Li, Joy C Yan, Eduardo M Sotomayor, Derek R Duckett, Bijal D Shah, Kenneth H Shain, Xiaohong Zhao, Jianguo Tao
ABSTRACT
Despite significant progress in the treatment of patients with diffuse large B-cell lymphoma (DLBCL) and mantle cell lymphoma (MCL), prognosis of patients with relapsed disease remains poor due to the emergence of drug resistance and subsequent disease progression. Identification of novel targets and therapeutic strategies for these diseases represents an urgent need. Here, we report that both MCL and DLBCL are exquisitely sensitive to transcription-targeting drugs, particular to THZ531, a covalent inhibitor of cyclin-dependent kinase 12 (CDK12). By implementing pharmacogenomics and a cell-based drug screen, we found that THZ531 leads to inhibition of oncogenic transcriptional programs, especially the DNA damage response (DDR) pathway, MYC target genes and the mTOR-4EBP1-MCL-1 axis, contributing to dramatic lymphoma suppression in vitro. We also identified de novo and established acquired THZ531 resistant lymphoma cells conferred by over-activation of the MEKERK and PI3K-AKT-mTOR pathways and upregulation of multidrug resistance-1 (MDR1) protein. Of note, EZH2 inhibitors reversed THZ531 resistance by competitive inhibition of MDR1 and, in combination with THZ531, synergistically inhibited MCL and DLBCL growth in vitro. Our study indicates that CDK12 inhibitor, alone or together with EZH2 inhibitors, offer promise as novel effective approaches for difficult to treat DLBCL and MCL.
Sigma-Aldrich
Anti-RNA polymerase II subunit B1 (phospho-CTD Ser-7) Antibody, clone 4E12, culture supernatant, clone 4E12, from rat
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Anti-RNA polymerase II subunit B1 (phospho-CTD Ser-5) Antibody, clone 3E8, culture supernatant, clone 3E8, from rat
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Anti-RNA polymerase II subunit B1 (phospho CTD Ser-2), clone 3E10 (rat monoclonal)., culture supernatant, clone 3E10, from rat