Merck
Probing the mechanical architecture of the vertebrate meiotic spindle.
Nature methods (2009-01-20)
Takeshi Itabashi, Jun Takagi, Yuta Shimamoto, Hiroaki Onoe, Kenta Kuwana, Isao Shimoyama, Jedidiah Gaetz, Tarun M Kapoor, Shin'ichi Ishiwata
ABSTRACT
Accurate chromosome segregation during meiosis depends on the assembly of a microtubule-based spindle of proper shape and size. Current models for spindle-size control focus on reaction diffusion-based chemical regulation and balance in activities of motor proteins. Although several molecular perturbations have been used to test these models, controlled mechanical perturbations have not been possible. Here we report a piezoresistive dual cantilever-based system to test models for spindle-size control and examine the mechanical features, such as deformability and stiffness, of the vertebrate meiotic spindle. We found that meiotic spindles prepared in Xenopus laevis egg extracts were viscoelastic and recovered their original shape in response to small compression. Larger compression resulted in plastic deformation, but the spindle adapted to this change, establishing a stable mechanical architecture at different sizes. The technique we describe here may also be useful for examining the micromechanics of other cellular organelles.
Sigma-Aldrich
Mineral oil, light oil, (neat)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Mineral oil, BioReagent, for molecular biology, light oil
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Mineral oil, meets USP testing specifications
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Mineral oil, suitable for preparation of Nujol mulls for infrared spectroscopy, light oil
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Mineral oil, PCR Reagent
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Mineral oil, light oil, suitable for mouse embryo cell culture, BioXtra
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Mineral oil, BioUltra, for molecular biology
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Mineral oil, heavy
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Mineral oil, light