Skip to Content
MilliporeSigma
Cyantraniliprole PESTANAL®, analytical standard

Cyantraniliprole

Cyantraniliprole PESTANAL®, analytical standard
Synonym(s):
3-Bromo-1-(3-chloro-2-pyridyl)-4′-cyano-2′-methyl-6′-(methylcarbamoyl)pyrazole-5-carboxanilide
Empirical Formula (Hill Notation):
C19H14BrClN6O2
CAS Number:
Molecular Weight:
473.71

Products

Product Category
Brand
Formula Weight
Manufacturer
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing