Skip to Content
Merck
3,4-Diethoxy-3-cyclobutene-1,2-dione 98%

3,4-Diethoxy-3-cyclobutene-1,2-dione

3,4-Diethoxy-3-cyclobutene-1,2-dione 98%
Synonym(s):
Diethyl squarate, Squaric acid diethyl ester
Linear Formula:
(C2H5O)2C4(=O)2
CAS Number:
Molecular Weight:
170.16

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing