PHL89524
Coptisine chloride
phyproof® Reference Substance
Synonym(s):
6,7-Dihydrobis[1,3]benzodioxolo[5,6-a:4‘,5‘-g]quinolizinium chloride, Coptisine hydrochloride
About This Item
Recommended Products
grade
primary reference standard
product line
phyproof® Reference Substance
Assay
≥95.0% (HPLC)
form
solid
manufacturer/tradename
PhytoLab
storage temp.
2-8°C
SMILES string
[Cl-].[n+]21c(cc5c(c6c(cc5)OCO6)c2)c3c(cc4c(c3)OCO4)CC1
InChI
1S/C19H14NO4.ClH/c1-2-16-19(24-10-21-16)14-8-20-4-3-12-6-17-18(23-9-22-17)7-13(12)15(20)5-11(1)14;/h1-2,5-8H,3-4,9-10H2;1H/q+1;/p-1
InChI key
LUXPUVKJHVUJAV-UHFFFAOYSA-M
Looking for similar products? Visit Product Comparison Guide
General description
Legal Information
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 2 Inhalation - Acute Tox. 2 Oral
Storage Class Code
6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service