Skip to Content
Merck
All Photos(1)

Documents

95661

Supelco

1,10-Diaminodecane

analytical standard

Synonym(s):

1,10-Decanediamine, Decamethylenediamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH2(CH2)10NH2
CAS Number:
Molecular Weight:
172.31
Beilstein:
1738591
MDL number:
UNSPSC Code:
41116107

grade

analytical standard

Quality Level

Assay

≥95.0% (GC)

technique(s)

HPLC: suitable

bp

140 °C/12 mmHg (lit.)

mp

59-61 °C (lit.)

format

neat

SMILES string

NCCCCCCCCCCN

InChI

1S/C10H24N2/c11-9-7-5-3-1-2-4-6-8-10-12/h1-12H2

InChI key

YQLZOAVZWJBZSY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service