Skip to Content
Merck
All Photos(1)

Key Documents

817392

TSKgel® GMHHR-M (5 µm) HPLC Columns

L x I.D. 30 cm × 7.8 mm, polystyrene-divinylbenzene phase

Synonym(s):

TSKgel® HHR Column

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700
eCl@ss:
32110501

Product Name

TSKgel® GMHHR-M HPLC Column, phase polystyrene-divinylbenzene, L × I.D. 30 cm × 7.8 mm, 5 μm particle size

material

stainless steel

Agency

suitable for USP L21

product line

TSKgel®

manufacturer/tradename

Tosoh 17392

parameter

0.5-1.0 mL/min standard flow rate
50 bar max. pressure
80 °C temperature

technique(s)

HPLC: suitable

L × I.D.

30 cm × 7.8 mm

matrix

polymer particle platform

matrix active group

polystyrene-divinylbenzene phase

particle size

5 μm

pore size

mixed

separation technique

size exclusion (SEC)

Looking for similar products? Visit Product Comparison Guide

General description

TSKgel GMHHR-M mixed bed columns are packed with rigid, multiple pore size, polymer beads. Mixed bed columns are commonly used for unknown samples and for the analysis of polymers with a broad molecular weight distribution. TSKgel GMHHR-M columns are shipped in THF, which can be exchanged for a wide variety of organic solvents. TSKgel GMHHR-M columns are available in 5 and 13μm particle sizes. We recommend the use of TSKgel GMHHR-M(S) (13μm) to prevent or limit shearing of ultra high molecular weight polymers, if this occurs when using small (5μm) particle size columns.
The TSKgel HHR column line consists of eight columns with different pore sizes, TSKgel G1000HHR through TSKgel G7000HHR, and five columns with an extended linear range of the calibration curve. All TSKgel HHR-type columns are shipped in THF, which can be exchanged for a much broader range of organic solvents than TSKgel HXL columns, although Hxl columns are more inert. The maximum operating temperature for most TSKgel HHR-type columns is 140°C, except for GMHHR columns, which can only be used up to 80°C. TSKgel HHR-type columns are conventional GPC columns with dimensions of 7.8mm ID x 30cm.

Physical form

Shipped in THF.

Other Notes

THF can be replaced with acetone, ethanol, quinoline, benzene, o-dichlorobenzene, ethyl acetate, dodecane, FC-113, carbon tetrachloride, dichloromethane, dichloroethane, trichloroethane, n-hexane, cyclohexane, xylene, tetrahydrofuran, chloroform, 1,4-dioxane, hexafluoroisopropanol, toluene, 1-chloronaphthalene, dimethylaceto acetamide, methyl ethyl ketone, trichlorobenzene, m-cresol, dimethylformamide, methylpyrrolidone, o-chlorophenol/chloroform, dimethyl sulfoxide, pyridine

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

TSKgel is a registered trademark of Tosoh Corporation

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service