Skip to Content
Merck
All Photos(1)

Key Documents

68137

Supelco

N-Nitrosodiethanolamine

analytical standard

Synonym(s):

2,2′-(Nitrosoimino)bisethanol, N,N-Diethanolnitrosamine, Bis(2-hydroxyethyl)nitrosamine, NDELA

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H10N2O3
CAS Number:
Molecular Weight:
134.13
Beilstein:
1762620
MDL number:
UNSPSC Code:
12352116
NACRES:
NA.05

grade

analytical standard

Quality Level

Assay

≥95%

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

format

neat

storage temp.

2-8°C

SMILES string

OCCN(N=O)CCO

InChI

1S/C4H10N2O3/c7-3-1-6(5-9)2-4-8/h7-8H,1-4H2

InChI key

YFCDLVPYFMHRQZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

FlameHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 1B - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

52.9 °F - closed cup

Flash Point(C)

11.6 °C - closed cup


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service