Skip to Content
Merck
All Photos(1)

Key Documents

46856-U

Supelco

EPA 531.1 Carbamate Mix

100 μg/mL each component in methanol, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
77101502
NACRES:
NA.24
Pricing and availability is not currently available.

grade

analytical standard

Quality Level

Agency

EPA 531.1

CofA

current certificate can be downloaded

feature

standard type calibration

packaging

ampule of 1 mL

concentration

100 μg/mL each component in methanol

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

multi-component solution

storage temp.

2-8°C

General description

EPA 531.1 Carbamate Mix is a multi-component solution containing certified reference materials (CRMs) such as aldicarb, aldicarb-sulfone, aldicarb-sulfoxide, carbofuran, carbofuran-3-hydroxy, 1-naphthyl-N-methylcarbamate, methiocarb, methomyl, oxamyl, and propoxur. It may be used for identifying and quantifying carbamates in drinking water. The pre-requisite of EPA 531.1 method is the use of gas chromatography (GC) with a selective detector, gas chromatography/mass spectrometry (GC/MS) or high-performance liquid chromatography (HPLC).

Application

EPA 531.1 Carbamate Mix may be used as a CRM standard to quantify the carbamate pesticides in fresh herbs by gas chromatography-mass spectrometry (GC-MS).[1]
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Analyte

Description

    Aldicarb
    Aldicarb-sulfone
    Aldicarb-sulfoxide
    Carbofuran
    Carbofuran-3-hydroxy
    1-Naphthyl-N-methylcarbamate
    Methiocarb
    Methomyl
    Oxamyl
    Propoxur
    See All (10)

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Chronic 3 - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

51.8 °F

Flash Point(C)

11 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Application of QuEChERS method for simultaneous determination of pesticide residues and PAHs in fresh herbs
Sadowska-Rociek A, et al.
Bulletin of Environmental Contamination and Toxicology, 90(4) (2013)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service