Skip to Content
Merck
All Photos(15)

Key Documents

ZRB06599

Sigma-Aldrich

Anti-Acetyl Histone H3 (Lys9 and 14) Antibody, clone 5H20, ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

14, 14Ac, H3/g, H3/t, H3K9, H3t, Histone H3 Ac Lysine 9, Histone H3.1t

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

rabbit (recombinant)

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

5H20, recombinant monoclonal
monoclonal

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 15.51 kDa
observed mol wt ~16 kDa

species reactivity

human

species reactivity (predicted by homology)

mouse, rat

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

immunocytochemistry: 1:100-1:1,000
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable using 1:100
inhibition assay: suitable using 1:1,000 (peptide)
western blot: suitable using 1:1,000

isotype

IgG

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

acetylation (Lys9, Lys14)

Gene Information

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb antibodies represent an entirely new generation of recombinant monoclonal antibodies.

Each ZooMAb antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb antibodies are reliably available and ready to ship when you need them.

Learn more about ZooMAb here.

Specificity

Clone 5H20 is a ZooMAb rabbit recombinant monoclonal antibody that specifically detects Histone H3 acetylated on lysines 9 and 14.

Immunogen

KLH-conjugated linear peptide corresponding to the first 21 amino acids (without methionine) surrounding acetylated lysines 9 and 14 of human Histone H3.

Application

Anti-Acetyl Histone H3 (Lys9 and 14), clone 5H20, ZooMAb, Cat. No. ZRB06599, is a recombinant Rabbit Monoclonal Antibody that specifically targets Histone H3 acetylated on Lysines 9 and 14 and has been tested for use in Immunocytochemistry, Immunohistochemistry (Paraffin), Peptide Inhibition, & WB.
Immunocytochemistry Analysis: A 1:1,000 dilution from a representative lot detected Acetyl Histone H3 (Lys9 and 14) in HeLa cells treated with Trichostatin A (TSA).

Immunohistochemistry (Paraffin) Analysis: A 1:100 dilution from a representative lot detected Acetyl Histone H3 (Lys9 and 14) in human breast cancer tissue sections.

Peptide Inhibition Analysis: A 1:1,000 dilution from a representative lot was used with HeLa cells treated with Sodium Butyrate for peptide block analysis.

Target description

Histone H3.1t (UniProt: Q16695; also known as H3/t, H3t, H3/g) is encoded by the HIST3H3 (also known as H3FT) gene (Gene ID: 8290) in human. Histones are highly conserved basic nuclear proteins that are responsible for the nucleosome structure of chromatin in eukaryotes. They play a central role in transcription regulation, DNA repair, DNA replication and chromosomal stability. DNA accessibility is regulated via a complex set of post-translational modifications of histones, also called histone code, and nucleosome remodeling. Two molecules of each of the four core histones (H2A, H2B, H3, and H4) form an octamer, around which DNA is wrapped in repeating units, called nucleosomes, which limits DNA accessibility to the cellular machineries that require DNA as a template. Histone H3 features a main globular domain and a long N-terminal tail, which protrudes from the globular nucleosome core and can undergo several different types of epigenetic modifications that influence cellular processes. Acetylation of histone H3 occurs at several different lysine positions in the histone tail and is performed by a family histone acetyl transferases. Histone H3 can also undergo methylation as well as acetylation on lysine 9. Acetylation on lysine 9 (H3K9ac) is reported to impair methylation at arginine 9 (H3R8me2s). A balance between acetylation and methylation is important for the establishment of specific chromatin structures. Acetylation of lysine 14 (H3K14Ac) is commonly seen in genes that are being actively transcribed into RNA. Histone H3 is reported to be hyperacetylated at lysines 9 and 14 following UV irradiation. Studies have shown that nucleosomes with H3K14ac modification have a higher affinity for purified chromatin remodeling complex RSC (Remodels the Structure of Chromatin) and by anchoring RSC, H3K14Ac plays an important role in the unfolding of strongly positioned nucleosomes during repair of UV damage. This ZooMAb recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals. (Ref.: Nicolas, E., et al. (2003). Mol. Cell. Biol. 23(5); 1614-1622; Duan, MR., and Smerdon, MJ (2014). J. Biol. Chem. 289(12); 8353-8363).

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS with 5% Trehalose, normal appearance a coarse or translucent resin. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 μL.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 μL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Vasileios Kampanis et al.
Cells, 10(1) (2021-01-01)
The central nervous system (CNS) does not recover from traumatic axonal injury, but the peripheral nervous system (PNS) does. We hypothesize that this fundamental difference in regenerative capacity may be based upon the absence of stimulatory mechanical forces in the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service