Skip to Content
Merck
All Photos(1)

Key Documents

Z748021

Reducing part for Hettich® swing-out rotor No. 1624

for 1 x 50 mL tubes (34 x 100 mm size)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41103911
NACRES:
NB.22

feature

Holds 50 mL tubes

packaging

pack of 1 ea

manufacturer/tradename

Hettich 1331

technique(s)

centrifugation: suitable

compatibility

for use with 1481, 1490, 1495 (buckets)

Legal Information

Hettich is a registered trademark of Andreas Hettich GmbH & Co. KG

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service