Z743586
Laser Tough-Spots®
diam. 7/16 in., white, pack of 4680 (234 labels per sheet)
Synonym(s):
solvent resistant labels, temperature resistant labels
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
UNSPSC Code:
41103032
NACRES:
NB.35
Recommended Products
packaging
pack of 4680 (234 labels per sheet)
manufacturer/tradename
Diversified Biotech SPOT-5000
diam.
7/16 in.
color
white
Looking for similar products? Visit Product Comparison Guide
General description
Tough-Spots are chemically inert, polyvinyl labels that adhere to all plastics. Tough-Spots withstand boiling water baths, conventional and cryogenic freezer temperatures, most organic solvents, caustic agents and other challenges without peeling, becoming brittle or degrading. Labels can be printed directly from the laser printer. 20 sheets per pack.
Legal Information
Tough-Spots is a registered trademark of Diversified Biotech
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service