Key Documents
Z719404
Sorenson low binding aerosol barrier tips
MultiGuard E, maximum volume 10 μL
Synonym(s):
aerosol barrier tips, barrier filter pipette tip, barrier filter tips, barrier tip, filter pipette tip, filter tip, low bind tip, pipet tip
About This Item
UNSPSC Code:
41121601
NACRES:
NB.01
manufacturer/tradename:
Sorenson 15020T
maximum volume:
10 μL
sterility:
sterile
feature:
DNase free
RNase free
barrier
packaging:
pack of 96 ea (Racks)
material
colorless
polypropylene
sterility
sterile
feature
DNase free
RNase free
barrier
packaging
pack of 96 ea (Racks)
manufacturer/tradename
Sorenson 15020T
maximum volume
10 μL
binding type
low binding surface
General description
Tips feature a new surface technology designed for quantitative assays in which sample binding can significantly alter results or for critical small volume protocols that require a high degree of accuracy. Ideal for molecular biology applications, such as PCR. Inert hydrophobic barrier prevents pipettor contamination.
Legal Information
Sorenson is a trademark of Sorenson BioScience, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)