Z325473
BRAND® SILBERBRAND ETERNA graduated pipette, calibrated to deliver (TD, EX)
capacity 2 mL
All Photos(1)
About This Item
Recommended Products
material
AR-Glass
feature
class B
graduation
subdivision 0.01 mL
supplementary to DIN range
packaging
pack of 12
manufacturer/tradename
BRAND 27072
capacity
2 mL
General description
AR-glass®. Graduated pipettes according to DIN EN ISO 835. Type 3 (zero point at the top), total delivery. Graduations and inscriptions in ETERNA amber stain, particularly resistant to aggressive cleaning methods.
Other Notes
BR27072 (India)
Legal Information
Araldite is a registered trademark of Huntsman Advanced Materials Inc.
BRAND is a registered trademark of BRAND GMBH + CO KG
Protocols
This page contains detailed information on working with and handling pipettes and pipetting aids.