Skip to Content
Merck
All Photos(2)

Key Documents

WH0007421M2

Sigma-Aldrich

Monoclonal Anti-VDR antibody produced in mouse

clone 2F4, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-NR1I1, Anti-vitamin D (1,25- dihydroxyvitamin D3) receptor

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

2F4, monoclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
indirect ELISA: suitable

isotype

IgG2aκ

GenBank accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... VDR(7421)

General description

VDR (vitamin D receptor) or vitamin D3 receptor is a member of the steroid and thyroid hormone receptors superfamily of trans-acting transcriptional regulatory factors. It is an intracellular polypeptide of 50-60kDa. This receptor is composed of a highly conserved cysteine, lysine, and arginine-rich DNA-binding domain, and a hydrophobic ligand-binding domain in its C-terminal. It shares size and sequence similarity to thyroid hormone receptor. VDR gene is localized to human chromosome 12q12–q14.

Immunogen

VDR (AAH60832, 1 a.a. ~ 427 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
MEAMAASTSLPDPGDFDRNVPRICGVCGDRATGFHFNAMTCEGCKGFFRRSMKRKALFTCPFNGDCRITKDNRRHCQACRLKRCVDIGMMKEFILTDEEVQRKREMILKRKEEEALKDSLRPKLSEEQQRIIAILLDAHHKTYDPTYSDFCQFRPPVRVNDGGGSHPSRPNSRHTPSFSGDSSSSCSDHCITSSDMMDSSSFSNLDLSEEDSDDPSVTLELSQLSMLPHLADLVSYSIQKVIGFAKMIPGFRDLTSEDQIVLLKSSAIEVIMLRSNESFTMDDMSWTCGNQDYKYRVSDVTKAGHSLELIEPLIKFQVGLKKLNLHEEEHVLLMAICIVSPDRPGVQDAALIEAIQDRLSNTLQTYIRCRHPPPGSHLLYAKMIQKLADLRSLNEEHSKQYRCLSFQPECSMKLTPLVLEVFGNEIS

Biochem/physiol Actions

VDR (vitamin D receptor) binds with its ligand 1a, 25-dihydroxyvitamin D3 [1,25(OH)2D3] or vitamin D3, and act upon the target cell nuclei to induce various biological effects. Vitamin D3 is also thought to have immunomodulatory effects, and gene polymorphisms are linked with pertussis. This variant might thus, affect the immune response and clinical outcome of Bordetella pertussis infection.

Physical form

Solution in phosphate buffered saline, pH 7.4

Legal Information

GenBank is a registered trademark of United States Department of Health and Human Services

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cloning and expression of full-length cDNA encoding human vitamin D receptor.
Baker AR
Proceedings of the National Academy of Sciences of the USA (1988)
Association Between VDR FokI Polymorphism and Intervertebral Disk Degeneration.
Zhao J
Genomics, Proteomics & Bioinformatics (2015)
Cloning and expression of full-length cDNA encoding human vitamin D receptor.
Han WG
PLoS ONE (2016)
Isabel Heidegger et al.
Molecular cancer, 21(1), 132-132 (2022-06-19)
Crosstalk between neoplastic and stromal cells fosters prostate cancer (PCa) progression and dissemination. Insight in cell-to-cell communication networks provides new therapeutic avenues to mold processes that contribute to PCa tumor microenvironment (TME) alterations. Here we performed a detailed characterization of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service