Skip to Content
Merck
All Photos(6)

Key Documents

WH0002643M1

Sigma-Aldrich

Monoclonal Anti-GCH1 antibody produced in mouse

clone 4A12, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-DYT5, Anti-GCH, Anti-GTP cyclohydrolase 1 (dopa-responsive dystonia), Anti-GTPCH1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

4A12, monoclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
immunoprecipitation (IP): suitable
indirect ELISA: suitable
western blot: 1-5 μg/mL

isotype

IgG2aκ

GenBank accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... GCH1(2643)

General description

Guanosine triphosphate (GTP) cyclohydrolase I (GCH1), a homodecameric protein belongs to the GTP cyclohydrolase family. Many isoforms of GCH1 are present due to alternative splicing. However, not all variants encode a functional enzyme. The GCH1 gene is mapped to human chromosome 14q22.2.

Immunogen

GCH1 (NP_000152, 84 a.a. ~ 172 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
ENPQRQGLLKTPWRAASAMQFFTKGYQETISDVLNDAIFDEDHDEMVIVKDIDMFSMCEHHLVPFVGKVHIGYLPNKQVLGLSKLARIV

Biochem/physiol Actions

Guanosine triphosphate (GTP) cyclohydrolase I (GCH1) catalyzes the synthesis of 7,8-dihydroneopterin triphosphate from GTP. It acts as a rate-limiting enzyme in the tetrahydrobiopterin (BH4) biosynthesis pathway. GCH1 requires zinc (Zn2+) for its catalytic function. It is crucial for dopamine synthesis, and its variants may have a role in the pathogenesis of Parkinson′s disease (PD). Mutations in the GCH1 gene are implicated in Segawa disease, hyperphenylalaninemia, and dopa-responsive dystonia (DRD). GCH1 gene polymorphisms are correlated to the pathophysiology of obstructive sleep apnea (OSA) and in neuropathic pain and stroke.

Physical form

Solution in phosphate buffered saline, pH 7.4

Legal Information

GenBank is a registered trademark of United States Department of Health and Human Services

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Samaneh Sheikhi Kouhsar et al.
Scientific reports, 9(1), 18664-18664 (2019-12-11)
Several studies have recently investigated the contribution of genetic factors in obstructive sleep apnea (OSA). Patients with OSA suffer from a reduction in nitric oxide (NO) serum level. This study investigated rs841, A930G p22phox, and rs1799983 polymorphisms in three critical
Takahiro Suzuki et al.
European journal of biochemistry, 271(2), 349-355 (2004-01-14)
GTP cyclohydrolase I (GCH) is the rate-limiting enzyme for the synthesis of tetrahydrobiopterin and its activity is important in the regulation of monoamine neurotransmitters such as dopamine, norepinephrine and serotonin. We have studied the action of divalent cations on the
Uladzislau Rudakou et al.
Neurobiology of aging, 73, 231-231 (2018-10-14)
GCH1 encodes the enzyme guanosine triphospahte (GTP) cyclohydrolase 1, essential for dopamine synthesis in nigrostriatal cells, and rare mutations in GCH1 may lead to Dopa-responsive dystonia (DRD). While GCH1 is implicated in genomewide association studies in Parkinson's disease (PD), only
Wai-Kwan Siu
Translational pediatrics, 4(2), 175-180 (2016-02-03)
The monoamine neurotransmitter disorders are a heterogeneous group of inherited neurological disorders involving defects in the metabolism of dopamine, norepinephrine, epinephrine and serotonin. The inheritance of these disorders is mostly autosomal recessive. The neurological symptoms are primarily attributable to cerebral

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service