Skip to Content
Merck
All Photos(1)

Key Documents

TRUEDEXF

Sigma-Aldrich

TrueGel3D Polymer

Modified for fast gelling (FAST-DEXTRAN),allow cell recovery

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352207
NACRES:
NA.71

shipped in

dry ice

storage temp.

−70°C

Looking for similar products? Visit Product Comparison Guide

General description

Dextrose polymers are functionalized with thiol-reactive maleimide groups, which react rapidly with thiol groups of crosslinkers to form hydrogels in few seconds. The thiol-reactive maleimide groups are also compatible to conjugate polymers with bioactive factors. The thioglycerol is used to maintain hydrogel strength and also as a control to compare cellular behavior in peptide-modified versus unmodified hydrogels. The buffers (pH7.2 and pH 5.5) are used to control pH and osmotic conditions to optimize time for gelation.

TrueGel3D enzymatic cell recovery solution (TRUEENZ-500UL) is used to dissolve dextran polymers for easy recovery of chemically fixed or live cells for post-culture analyses including RT-PCR, western blots and other molecular biological studies.

Application

FAST-DEXTRAN polymer is used with crosslinkers (CD/PEG crosslinkers) to establish stable hydrogels

Features and Benefits

  • Contains dextrose polymer functionalized with maleimide groups for quick gelation
  • Degradable polymer

Components

TRUEDEXF contains FAST-DEXTRAN lyophilized with reconstitution buffer, TrueGel3D thioglycerol and water

Caution

Avoid frequent freeze-thaw cycles and store FAST-DEXTRAN in small portions

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Met. Corr. 1 - Skin Irrit. 2

Storage Class Code

8A - Combustible corrosive hazardous materials


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Troubleshoot TrueGel3D™ hydrogel formation and cell viability issues with RGD-degradable polymer and CD cell-degradable crosslinker.

Troubleshoot TrueGel3D™ hydrogel formation and cell viability issues with RGD-degradable polymer and CD cell-degradable crosslinker.

Troubleshoot TrueGel3D™ hydrogel formation and cell viability issues with RGD-degradable polymer and CD cell-degradable crosslinker.

Troubleshoot TrueGel3D™ hydrogel formation and cell viability issues with RGD-degradable polymer and CD cell-degradable crosslinker.

See All

Protocols

An advanced protocol is available for TrueGel 3D kits for 3D cell culture for peptides concentration or stiffness optimisation

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service