Skip to Content
Merck
All Photos(1)

Key Documents

SRP3142

Sigma-Aldrich

PF-4 (CXCL4) human

recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture

Synonym(s):

CXCL4, Ironplact, Oncostatin A, Platelet Factor-4

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

biological source

human

recombinant

expressed in E. coli

Assay

≥98% (HPLC)
≥98% (SDS-PAGE)

form

lyophilized

potency

1.0-10.0 ng/mL

mol wt

7.8 kDa

packaging

pkg of 20 μg

technique(s)

cell culture | mammalian: suitable

impurities

<0.1 EU/μg endotoxin, tested

color

white

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... PF4(5196)

General description

Platelet factor-4 (PF-4) is a CXC chemokine that is expressed in megakaryocytes and stored in the alpha-granules of platelets. The gene encoding this protein is localized on human chromosome 4. Recombinant human PF-4 is a 7.8kDa protein containing 70 amino acid residues, including the four highly conserved residues present in CXC chemokines.

Biochem/physiol Actions

Platelet factor-4 (PF-4) is chemotactic towards neutrophils and monocytes and has been shown to inhibit angiogenesis. This protein associates with heparin and neutralizes it. It also neutralizes various negatively charged proteoglycans.

Sequence

EAEEDGDLQC LCVKTTSQVR PRHITSLEVI KAGPHCPTAQ LIATLKNGRK ICLDLQAPLY KKIIKKLLES

Physical form

Lyophilized with no additives.

Reconstitution

Centrifuge the vial prior to opening. Reconstitute in water to a concentration of 0.1-1.0 mg/ml. Do not vortex. This solution can be stored at 2-8°C for up to 1 week. For extended storage, it is recommended to further dilute in a buffer containing a carrier protein (example 0.1% BSA) and store in working aliquots at -20°C to -80°C.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chemokines and Chemokine Receptors
Manel Juan
Wiley Guide to Chemical Incompatibilities (2013)
S S Huang et al.
The Journal of biological chemistry, 257(19), 11546-11550 (1982-10-10)
A large scale purification procedure for the human platelet factor 4 proteoglycan carrier molecule has been developed. A yield of 46% and a 33,000-fold purification have been achieved, using poly-L-lysine-Sepharose affinity column chromatography, PF4-agarose affinity column chromatography, and Bio-Gel A-0.5m
CXCL4 Plasma Levels Are Not Associated with the Extent of Coronary Artery Disease or with Coronary Plaque Morphology.
Erbel C
PLoS ONE, 10(11), e0141693-e0141693 (2015)
B Scheuerer et al.
Blood, 95(4), 1158-1166 (2000-02-09)
Unstimulated monocytes rapidly undergo physiological changes resulting in programmed cell death (apoptosis) while stimuli promoting differentiation of these cells into macrophages were shown to inhibit apoptotic processes. In the present study, we report that the platelet-derived alpha-chemokine platelet factor 4
Yong Du et al.
The Journal of experimental medicine, 219(7) (2022-06-01)
Chemokines control the migratory patterns and positioning of immune cells to organize immune responses to pathogens. However, many chemokines have been associated with systemic autoimmune diseases that have chronic IFN signatures. We report that a series of chemokines, including CXCL4

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service