Skip to Content
Merck
All Photos(1)

Key Documents

SRP3067

Sigma-Aldrich

IGF-Binding Protein-3 human

recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture

Synonym(s):

Growth-hormone-dependant binding protein, IBP-3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
51111800
NACRES:
NA.32

biological source

human

recombinant

expressed in E. coli

Assay

≥98% (HPLC)
≥98% (SDS-PAGE)

form

lyophilized

potency

≤0.2 μg/mL

mol wt

28.8 kDa

packaging

pkg of 25 μg

technique(s)

cell culture | mammalian: suitable

impurities

<0.1 EU/μg endotoxin, tested

color

white to off-white

suitability

suitable for molecular biology

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... IGFBP3(3486)

Related Categories

General description

Research area: Cell Signaling

IGFBPs (insulin-like growth factor-binding proteins) are mainly required to regulate IGFs (insulin like growth factors) activities. IGFBP3 is a multifunctional protein and is secreted by various cells. It is most ample circulating IGFBP and works in IGF-dependent and -independent mechanisms. The IGFBP3 gene is mapped to human chromosome 7p12.3.

Recombinant human IGF-BP3 is a 28.8kDa protein consisting of 264 amino acid residues.

Application

IGF-Binding Protein-3 human has been used to study its association with follicular fluid and subsequent effect on embryo development.

Biochem/physiol Actions

IGFBP3 (insulin-like growth factor-binding protein 3) is involved in cell survival as well as cell death and tumorigenesis. Epigenetic changes in IGFBP3 affect non-small-cell lung cancer, hepatocellular carcinoma, ovarian cancer, skin cancer, urological cancers, breast cancer and gastric cancer. IGFBP3 is a primary growth inhibitor and can reverse the mitogenic functions of IGF-1 (insulin like growth factor 1) by inducing apoptosis.IGFBP-3 functions both in an IGF-dependent and independent manner. Its IGF-dependent roles involve endocrine function by delivering IGFs to target cells and activating downstream signaling. Meanwhile, its IGF-independent roles include interactions with extracellular matrix and plasma membrane proteins, as well as translocation into the cytoplasm and nucleus. The C-terminal domain has cell-penetrating abilities, enabling the generation of short peptides for intracellular delivery of drugs or genes.

Sequence

GASSGGLGPV VRCEPCDARA LAQCAPPPAV CAELVREPGC GCCLTCALSE GQPCGIYTER CGSGLRCQPS PDEARPLQAL LDGRGLCVNA SAVSRLRAYL LPAPPAPGNA SESEEDRSAG EVESPSVSST HRVSDPKFHP LHSKIIIIKK GHAKDSQRYK VDYESQSTDT QNFSSESKRE TEYGPCRREM EDTLNHLKFL NVLSPRGVHI PNCDKKGFYK KKQCRPSKGR KRGFCWCVDK YGQPLPGYTT KGKEDVHCYS MQSK

Physical form

Lyophilized from 20mM HCl.

Reconstitution

Centrifuge the vial prior to opening. Reconstitute in water to a concentration of 0.1-1.0 mg/ml. Do not vortex. This solution can be stored at 2-8°C for up to 1 week. For extended storage, it is recommended to further dilute in a buffer containing a carrier protein (example 0.1% BSA) and store in working aliquots at -20°C to -80°C.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Insulin-like growth factor binding protein-3 mediates interleukin-24-induced apoptosis through inhibition of the mTOR pathway in prostate cancer.
Du Y, et al.
Oncology Reports, 34, 2273-2273 (2015)
Evaluation of Insulin-like Growth Factor-1 and Insulin-like Growth Factor Binding Protein-3 Expression Levels in Patients with Chronic Lymphocytic Leukemia.
Ayer M, et al.
Turkish journal of haematology : official journal of Turkish Society of Haematology, 33, 335-335 (2016)
A genome-wide association study identifies novel loci associated with circulating IGF-I and IGFBP-3.
Kaplan RC, et al.
Human Molecular Genetics, 20, 1241-1241 (2011)
Shailly Varma Shrivastav et al.
Frontiers in cell and developmental biology, 8, 286-286 (2020-06-02)
Insulin-like growth factor (IGF) binding protein-3 (IGFBP-3), one of the six members of the IGFBP family, is a key protein in the IGF pathway. IGFBP-3 can function in an IGF-dependent as well as in an IGF-independent manner. The IGF-dependent roles
The low IGFBP-3 level is associated with esophageal cancer patients: a meta-analysis.
Song G, et al.
World Journal of Surgical Oncology, 14, 307-307 (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service