SRP3049
GDF-2 human
recombinant, expressed in CHO cells, ≥95% (SDS-PAGE), ≥95% (HPLC), suitable for cell culture
Synonym(s):
BMP-9, Growth/Differentiation Factor-2
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352202
NACRES:
NA.32
Recommended Products
biological source
human
recombinant
expressed in CHO cells
Assay
≥95% (HPLC)
≥95% (SDS-PAGE)
form
lyophilized
potency
0.5-1.9 ng/mL
mol wt
24.1 kDa
packaging
pkg of 10 μg
technique(s)
cell culture | mammalian: suitable
impurities
<0.1 EU/μg endotoxin, tested
color
white to off-white
UniProt accession no.
shipped in
wet ice
storage temp.
−20°C
Gene Information
human ... GDF2(2658)
General description
GDF-2 belongs to the TGF-β cytokine family whose members play an important role during prenatal development and postnatal growth, remodeling and maintenance of a variety of tissues and organs. GDF-2 is expressed mainly in nonparenchymal cells of the liver, but is also found in other various cells and tissues. GDF-2 can signal through the ALK1 receptor and has been implicated in a number of physiologic events. These include regulation of the hepatic reticuloendothelial system, glucose homeostasis, and iron homeostasis, as well as the inhibition of angiogenesis. Recombinant human GDF-2 is a 24.1 kDa disulfide linked homodimeric protein consisting of two 110 amino acid polypeptide chains.
Biochem/physiol Actions
GDF-2 belongs to the TGF-β cytokine family whose members play an important role during prenatal development and postnatal growth, remodeling and maintenance of a variety of tissues and organs. Recombinant human GDF-2 is a 24.1 kDa disulfide linked homodimeric protein consisting of two 110 amino acid polypeptide chains.
Sequence
SAGAGSHCQK TSLRVNFEDI GWDSWIIAPKE YEAYECKGGC FFPLADDVTPTK HAIVQTLVHL KFPTKVGKAC CVPTKLSPIS VLYKDDMGVP TLKYHYEGMS VAECGCR
Physical form
Lyophilized with no additives.
Reconstitution
Centrifuge the vial prior to opening. Reconstitute in water to a concentration of 0.1-1.0 mg/ml. Do not vortex. This solution can be stored at 2-8°C for up to 1 week. For extended storage, it is recommended to further dilute in a buffer containing a carrier protein (example 0.1% BSA) and store in working aliquots at -20°C to -80°C.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Monica A Brown et al.
The Journal of biological chemistry, 280(26), 25111-25118 (2005-04-27)
Bone morphogenetic proteins (BMPs), a subset of the transforming growth factor (TGF)-beta superfamily, regulate a diverse array of cellular functions during development and in the adult. BMP-9 (also known as growth and differentiation factor (GDF)-2) potently induces osteogenesis and chondrogenesis
Sandrine Levet et al.
Proceedings of the National Academy of Sciences of the United States of America, 112(25), E3207-E3215 (2015-06-10)
The transition to pulmonary respiration after birth requires rapid alterations in the structure of the mammalian cardiovascular system. One dramatic change that occurs is the closure of the ductus arteriosus (DA), an arterial connection in the fetus that directs blood
Ferdos Alaa El Din et al.
PloS one, 10(7), e0132111-e0132111 (2015-07-16)
Hereditary Hemorrhagic Telangiectasia syndrome (HHT) or Rendu-Osler-Weber (ROW) syndrome is an autosomal dominant vascular disorder. Two most common forms of HHT, HHT1 and HHT2, have been linked to mutations in the endoglin (ENG) and activin receptor-like kinase 1 (ACVRL1or ALK1)
Wen-Jia Ai et al.
Stem cells and development, 24(11), 1320-1331 (2015-01-21)
A low-efficiency yield hinders the use of stem cells as a source of endothelial cells (ECs) for therapeutic vascularization, and the diversity of the transforming growth factor-β (TGF-β) superfamily has undermined understanding the effects of its potent vascularization-inducing. Herein, we
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service