Skip to Content
Merck
All Photos(1)

Key Documents

SRP0691

Sigma-Aldrich

PI3K (P85 ALPHA) Protein

recombinant

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.77

recombinant

expressed in baculovirus infected Sf9 cells

Assay

70-95% (SDS-PAGE)

form

frozen liquid

mol wt

~86 kDa

concentration

0.2 μg/μL

accession no.

NM_181523

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... PI3K (5295)

Application

Useful for Western blot and kinase assays.

Biochem/physiol Actions

Recombinant full-length human PI3K (p85alpha) was expressed by Baculovirus in Sf9 insect cells using an N-terminal His tag. The PI3K comprises of a 110 kDa catalytic subunit and a 85 kDa regulatory subunit. A number of isoforms of the 110 kDa catalytic subunit and the 85 kDa regulatory subunit exist in cells. p85α modulates the interaction between PI3K and platelet-derived growth factor receptor (1). Furthermore, estrogen receptor isoform ER-alpha binds in a ligand-dependent manner to the p85-alpha regulatory subunit of PI3K. Stimulation with estrogen increases ER-alpha-associated PI3K activity, leading to the activation of protein kinase B/AKT and endothelial nitric oxide synthase (eNOS) (2).

Reconstitution

Formulated in 50mM sodium phosphate, pH 7.0, 300mM NaCl, 150mM imidazole, 0.1mM PMSF, 0.25mM DTT, with 25% glycerol.

Storage and Stability

Store in aliquots at -70°C. Avoid freeze/thaw cycles.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Repr. 1B - Skin Irrit. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service