Product SRP0519, Human PD-1 (CD279), FC Fusion is supplied in 8 mM phosphate pH 7.4, 110 mM NaCl, 2.2 mM KCl, and 20% glycerol.
SRP0519
Human PD-1 (CD279), FC Fusion
recombinant, expressed in HEK 293 cells
Synonym(s):
CD279, HPD-L, PD-1, PD1, PDCD1, Programmed cell death 1, SLEB2
About This Item
Recommended Products
recombinant
expressed in HEK 293 cells
form
frozen liquid
mol wt
42.6 kDa
UniProt accession no.
shipped in
dry ice
storage temp.
−70°C
Gene Information
human ... PDCD1(5133)
General description
Physical form
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2
Storage Class Code
10 - Combustible liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
-
Hi, I would like to know the buffer composition of this product. Thanks.
1 answer-
Helpful?
-
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service