SML3825
MM0299
≥98% (HPLC)
Synonym(s):
4,4a,5,5a,6,6a-Hexahydro-2-[4-[2-(4-methoxyphenyl)-2-oxoethoxy]phenyl]-4,6-ethenocycloprop[f]isoindole-1,3(2H,3aH)-dione
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C26H23NO5
CAS Number:
Molecular Weight:
429.46
MDL number:
UNSPSC Code:
51111800
UNSPSC Code:
12352200
NACRES:
NA.21
Recommended Products
Quality Level
Assay
≥98% (HPLC)
form
powder
color
white to beige
solubility
DMSO: 2 mg/mL, clear
storage temp.
-10 to -25°C
Biochem/physiol Actions
Cell penetrant, selective and potent inhibitor of lanosterol synthase (LSS) that exhibits anti-glioblastoma activity
MM0299 is a cell penetrant, selective and potent inhibitor of lanosterol synthase (LSS) that exhibits anti-glioblastoma activity. It potently inhibits Mut6 (Glioblastoma stem-like cells) cell growth by depleting cellular cholesterol. MM0299 stimulates 24(S),25-epoxycholesterol (EPC) synthesis in GBM tumors, which induces toxicity in glioma stem-like cells.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Selective and brain-penetrant lanosterol synthase inhibitors target glioma stem-like cells by inducing 24(S),25-epoxycholesterol production
Cell Chemical Biology, 30(2), 214-229 (2023)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service