Skip to Content
Merck
All Photos(1)

Key Documents

SML3823

Sigma-Aldrich

KCI807

≥98% (HPLC)

Synonym(s):

5,3’-Dihydroxyflavone, 5-Hydroxy-2-(3-hydroxyphenyl)-4H-1-benzopyran-4-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H10O4
CAS Number:
Molecular Weight:
254.24
MDL number:
UNSPSC Code:
51111800
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

0.2 mg/mL (DMSO:EtOH (2:1),clear, warmed, clear colorless solution)

storage temp.

room temp

SMILES string

[o]1c2c([c](cc1c3cc(ccc3)O)=O)c(ccc2)O

InChI key

SZMQXDYEGKRZDG-UHFFFAOYSA-N

Biochem/physiol Actions

Cell penetrant, selective and potent inhibitor of the androgen receptor (AR)-ELK1 interaction
KCI807 is a cell penetrant, selective and potent inhibitor of the androgen receptor (AR)-ELK1 interaction. KCI807 interrupts association of the androgen receptor (AR) with transcription activator ELK1 (ETS family transcription factor ELK1). It potently suppresses growth of enzalutamide-resistant prostate cancer tumor xenografts in rodents.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rayna Rosati et al.
Clinical cancer research : an official journal of the American Association for Cancer Research, 24(24), 6509-6522 (2018-09-07)
Testosterone suppression in prostate cancer is limited by serious side effects and resistance via restoration of androgen receptor (AR) functionality. ELK1 is required for AR-dependent growth in various hormone-dependent and castration-resistant prostate cancer models. The amino-terminal domain of AR docks
The Small Molecule Antagonist KCI807 Disrupts Association of the Amino-Terminal Domain of the Androgen Receptor with ELK1 by Modulating the Adjacent DNA Binding Domain
Molecular Pharmacology, 103(4), 211-220 (2023)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service