Skip to Content
Merck
All Photos(1)

Key Documents

SML3808

Sigma-Aldrich

Firategrast

≥98% (HPLC)

Synonym(s):

SB 683699, T 0047, T-0047, (αS)-α-[(2,6-Difluorobenzoyl)amino]-4′-(ethoxymethyl)-2′,6′-dimethoxy[1,1′-biphenyl]-4-propanoic acid, (S)-2-(2,6-Difluorobenzamido)-3-(4′-(ethoxymethyl)-2′,6′-dimethoxy-[1,1′-biphenyl]-4-yl)propanoic acid, SB-683699

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H27F2NO6
CAS Number:
Molecular Weight:
499.50
UNSPSC Code:
51111800
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

Biochem/physiol Actions

Firategrast is an orally available, potent and selective antagonist of α4β1/α4β7 integrins. Firategrast inhibits α4β1/α4β7 integrin-mediated transferring of lymphocytes into the central nervous system (CNS). In phase II trial it demonstrated a reduction of new contrast enhancing MRI lesions in patients with relapsing remitting MS.
Orally available, potent and selective antagonist of α4β1/α4β7 integrins

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Current and emerging disease-modulatory therapies and treatment targets for multiple sclerosis
Journal of Internal Medicine, 289(6), 771-791 (2021)
Comprehensive bioinformatic analysis reveals a cancer-associated fibroblast gene signature as a poor prognostic factor and potential therapeutic target in gastric cancer
BMC Cancer, 22(1), 692-692 (2022)
Firategrast for relapsing remitting multiple sclerosis: a phase 2, randomised, double-blind, placebo-controlled trial
Lancet Neurology, 11(2), 131-139 (2012)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service