Skip to Content
Merck
All Photos(1)

Key Documents

SML3774

Sigma-Aldrich

PITCOIN2

≥98% (HPLC)

Synonym(s):

2-[[3,4-Dihydro-4-oxo-3-(2-phenylethyl)-2-pteridinyl]thio]-N-[4-(3-hydroxyphenyl)-2-thiazolyl]acetamide, N-(4-(3-Hydroxyphenyl)thiazol-2-yl)-2-((4-oxo-3-phenethyl-3,4-dihydropteridin-2-yl)thio)acetamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H20N6O3S2
CAS Number:
Molecular Weight:
516.59
UNSPSC Code:
12352200
NACRES:
NA.21

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

Biochem/physiol Actions

PITCOIN2 is a cell permeable, potent, and highly selective inhibitor Phosphatidylinositol 3-kinase type 2α (PI3KC2α) catalytic activity. Also, PITCOIN2 is a moderate inhibitor of PI3KC2β. PITCOIN2 inhibits PI3KC2α-mediated synthesis of PI(3,4)P2 and PI(3)P. It exhibits potent antithrombotic activity by counteracting platelet membrane remodeling.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Development of selective inhibitors of phosphatidylinositol 3-kinase C2?
Nature Chemical Biology, 19(1), 18-27 (2023)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service