Skip to Content
Merck
All Photos(1)

Key Documents

SML3770

Sigma-Aldrich

BMS-466442

≥98% (HPLC)

Synonym(s):

(S)-Methyl 3-(1-benzyl-1H-imidazol-4-yl)-2-(6-benzyloxy-5-methoxy-1H-indole-2-carboxamido)propanoate, BMS 466442, BMS466442, N-[[5-Methoxy-6-(phenylmethoxy)-1H-indol-2-yl]carbonyl]-1-(phenylmethyl)-L-histidine methyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C31H30N4O5
CAS Number:
Molecular Weight:
538.59
MDL number:
UNSPSC Code:
12352200

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

-10 to -25°C

SMILES string

COC([C@H](CC1=CN(C=N1)CC2=CC=CC=C2)NC(C(NC3=C4)=CC3=CC(OC)=C4OCC5=CC=CC=C5)=O)=O

InChI key

UUCAHZCMRZOTNF-MHZLTWQESA-N

Biochem/physiol Actions

Potent and selective alanine serine cysteine transporter-1 (ASC-1; SLC7A10) inhibitor that effectively blocks ASC-1-mediated cellular D-Ser uptake.



BMS-466442 is a selective alanine serine cysteine transporter-1 (ASC-1) inhibitor (D-Ser uptake IC50 = 36.8 nM and 19.7 nM using HEK human ASC-1 transfectants or primary rat embryonic cortical cultures, respectively) with good selectivity over >40 other targets, including LAT-2 or ASCT-2 (IC50 >10 μM).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rini Arianti et al.
FEBS letters, 595(16), 2085-2098 (2021-07-02)
Brown and beige adipocytes dissipate energy by uncoupling protein 1 (UCP1)-dependent and UCP1-independent thermogenesis, which may be utilized to develop treatments against obesity. We have found that mRNA and protein expression of the alanine/serine/cysteine transporter-1 (ASC-1) was induced during adipocyte
Inhibition of the Alanine-Serine-Cysteine-1 Transporter by BMS-466442
ACS Chemical Neuroscience, 10(5), 2510-2517 (2019)
Jeffrey M Brown et al.
Journal of neurochemistry, 129(2), 275-283 (2013-11-26)
NMDA receptor hypofunction is hypothesized to contribute to cognitive deficits associated with schizophrenia. Since direct activation of NMDA receptors is associated with serious adverse effects, modulation of the NMDA co-agonists, glycine or D-serine, represents a viable alternative therapeutic approach. Indeed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service