Skip to Content
Merck
All Photos(1)

Key Documents

SML3696

Sigma-Aldrich

DHMEQ

≥98% (HPLC)

Synonym(s):

2-Hydroxy-N-[2-hydroxy-5-oxo-7-oxabicyclo[4.1.0]hept-3-en-3-yl]benzamide, DHM2EQ, Dehydroxymethylepoxyquinomicin, rel-2-Hydroxy-N-[(1R,2R,6R)-2-hydroxy-5-oxo-7-oxabicyclo[4.1.0]hept-3-en-3-yl]benzamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H11NO5
CAS Number:
Molecular Weight:
261.23
MDL number:
UNSPSC Code:
12352200

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

O[C@@H]1[C@@]2([H])[C@](C(C=C1NC(C3=C(C=CC=C3)O)=O)=O)([H])O2

InChI

1S/C13H11NO5/c15-8-4-2-1-3-6(8)13(18)14-7-5-9(16)11-12(19-11)10(7)17/h1-5,10-12,15,17H,(H,14,18)/t10-,11+,12-/m0/s1

InChI key

IUOMATKBBPCLFR-TUAOUCFPSA-N

Biochem/physiol Actions

Potent, selective, and irreversible inhibitor of NF-κB that covalently binds to a cysteine residue.



DHMEQ is a potent, selective, and irreversible inhibitor of NF-κB that covalently binds to a cysteine residue. It inhibits inflammatory cytokine expression in mice models. DHMEQ is a potent and non/low toxic in vivo anti-inflammatory agent effective in number of animal models.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jun Ma et al.
Cells, 10(9) (2021-09-29)
General inflammatory diseases include skin inflammation, rheumatoid arthritis, inflammatory bowel diseases, sepsis, arteriosclerosis, and asthma. Although these diseases have been extensively studied, most of them are still difficult to treat. Meanwhile, NF-κB is a transcription factor promoting the expression of
Reactive oxygen species trigger NF-?B-mediated NLRP3 inflammasome activation involvement in low-dose CdTe QDs exposure-induced hepatotoxicity
Redox Biology, 47, 102157-102157 (2021)
Dehydroxymethylepoxyquinomicin, a novel nuclear factor-?B inhibitor, prevents the development of cyclosporine A nephrotoxicity in a rat model
BMC Pharmacology & Toxicology, 21(1), 60-60 (2020)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service