Skip to Content
Merck
All Photos(1)

Key Documents

SML3481

Sigma-Aldrich

Norursodeoxycholic acid

≥98% (HPLC)

Synonym(s):

(3α,5β,7β)-3,7-Dihydroxy-24-norcholan-23-oic acid, 24-Norursodeoxycholic acid, 3α,7β-Dihydroxy-24-nor-5β-cholan-23-oic acid, Nourcholic acid, nor-UDCA, nor-Ursodeoxycholic acid, norUDCA

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H38O4
CAS Number:
Molecular Weight:
378.55
MDL number:
UNSPSC Code:
12352106
NACRES:
NA.77
Pricing and availability is not currently available.

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

-10 to -25°C

SMILES string

O[C@@H]1[C@@H]2[C@@H]([C@@]4([C@H](C1)C[C@@H](CC4)O)C)CC[C@]3([C@H]2CC[C@@H]3[C@@H](CC(=O)O)C)C

InChI

1S/C23H38O4/c1-13(10-20(26)27)16-4-5-17-21-18(7-9-23(16,17)3)22(2)8-6-15(24)11-14(22)12-19(21)25/h13-19,21,24-25H,4-12H2,1-3H3,(H,26,27)/t13-,14+,15-,16-,17+,18+,19+,21+,22+,23-/m1/s1

InChI key

QYYDXDSPYPOWRO-JHMCBHKWSA-N

Biochem/physiol Actions

Orally active bile acid, side chain-shortened ursodeoxycholic acid (UDCA) homologue with in vivo efficacy in cholestatic and fibrotic liver disease models.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service