Skip to Content
Merck
All Photos(1)

Key Documents

SML3263

Sigma-Aldrich

Edoxaban tosylate

≥98% (HPLC)

Synonym(s):

DU 176 tosylate, N-(5-Chloropyridin-2-yl)-N′-[(1S,2R,4S)-4-[(dimethylamino)carbonyl]-2-[[(5-methyl-4,5,6,7-tetrahydrothiazolo[5,4-c]pyridin-2-yl)carbonyl]amino]cyclohexyl]ethanediamide tosylate, N1-(5-Chloro-2-pyridinyl)-N2-[(1S,2R,4S)-4-[(dimethylamino)carbonyl]-2-[[(4,5,6,7-tetrahydro-5-methylthiazolo[5,4-c]pyridin-2-yl)carbonyl]amino]cyclohexyl]ethanediamide tosylate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H30ClN7O4S·C7H8O3S
CAS Number:
Molecular Weight:
720.26
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77
Pricing and availability is not currently available.

Quality Level

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

O=C(C(NC1=CC=C(Cl)C=N1)=O)N[C@H]2CC[C@H](C(N(C)C)=O)C[C@H]2NC(C3=NC4=C(S3)CN(C)CC4)=O.CC5=CC=C(C=C5)S(=O)(O)=O

InChI

1S/C24H30ClN7O4S.C7H8O3S/c1-31(2)24(36)13-4-6-15(27-20(33)21(34)30-19-7-5-14(25)11-26-19)17(10-13)28-22(35)23-29-16-8-9-32(3)12-18(16)37-23;1-6-2-4-7(5-3-6)11(8,9)10/h5,7,11,13,15,17H,4,6,8-10,12H2,1-3H3,(H,27,33)(H,28,35)(H,26,30,34);2-5H,1H3,(H,8,9,10)/t13-,15-,17+;/m0./s1

InChI key

ZLFZITWZOYXXAW-QXXZOGQOSA-N

Biochem/physiol Actions

Direct factor Xa inhibitor; antithrombotic anticoagulant
Edoxaban is a direct, orally available, selective, reversible and competitive inhibitor of human factor Xa.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service