Skip to Content
Merck
All Photos(1)

Key Documents

SML2837

Sigma-Aldrich

Tizoxanide

≥98% (HPLC)

Synonym(s):

2-Hydroxy-N-(5-nitro-2-thiazolyl)benzamide, 2-Hydroxy-N-(5-nitrothiazol-2-yl)benzamide, Desacetyl-NTZ, Desacetyl-nitazoxanide, NSC 697856, NTZ metabolite, Nitazoxanide metabolite, TIZ

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H7N3O4S
CAS Number:
Molecular Weight:
265.25
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

OC1=C(C(NC2=NC=C([N+]([O-])=O)S2)=O)C=CC=C1

InChI

1S/C10H7N3O4S/c14-7-4-2-1-3-6(7)9(15)12-10-11-5-8(18-10)13(16)17/h1-5,14H,(H,11,12,15)

InChI key

FDTZUTSGGSRHQF-UHFFFAOYSA-N

Biochem/physiol Actions

Tizoxanide (TIZ) is the active metabolite of the broad-spectrum parasiticidal drug nitazoxanide (NTZ), a noncompetitive inhibitor against bacterial pyruvate-ferredoxin oxidoreductase (PFOR Ki 2 to 10 μM). NTZ deacetylates rapidly to TIZ in plasma (t1/2 ~6 min at 37°C in human plasma) and in the presence of liver microsomes. NTZ is also reported to exhibit antiviral activity against Hepatitis C, Norovirus, Paramyxovirus, Influenza, Vaccinia, and Zika.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Z Zhao et al.
Journal of veterinary pharmacology and therapeutics, 33(2), 147-153 (2010-05-07)
The pharmacokinetics of tizoxanide (T), the active metabolite of nitazoxanide (NTZ), and its protein binding ability in goat plasma and in the solutions of albumin and alpha-1-acid-glycoprotein were investigated. The plasma and protein binding samples were analyzed using a high-performance
J Broekhuysen et al.
International journal of clinical pharmacology and therapeutics, 38(8), 387-394 (2000-09-13)
Nitazoxanide (N), a new broad-spectrum parasiticidal agent, is rapidly deacetylated to tizoxanide (T). The objective of the study was to determine if metabolites other than T are present in the plasma and excreted after single dose oral administration of radiocarbon-labelled
Paul S Hoffman et al.
Antimicrobial agents and chemotherapy, 51(3), 868-876 (2006-12-13)
Nitazoxanide (NTZ) exhibits broad-spectrum activity against anaerobic bacteria and parasites and the ulcer-causing pathogen Helicobacter pylori. Here we show that NTZ is a noncompetitive inhibitor (K(i), 2 to 10 microM) of the pyruvate:ferredoxin/flavodoxin oxidoreductases (PFORs) of Trichomonas vaginalis, Entamoeba histolytica
A Stockis et al.
International journal of clinical pharmacology and therapeutics, 40(5), 213-220 (2002-06-08)
Nitazoxanide (N) is a new broad-spectrum intestinal antiparasitic agent. Deacetyl-N or tizoxanide (T) and its glucuronide (TG) are the major circulating species metabolites after oral administration of N. Bioavailability is substantially increased by food. The objectives of this phase IA
Jinqiu Liu et al.
Biomedical chromatography : BMC, 30(11), 1744-1749 (2016-04-24)
Nitazoxanide (NTZ) is a broad-spectrum antimicrobial agent. Tizoxanide (T) and tizoxanide glucuronide (TG) are the major circulating metabolites after oral administration of NTZ. A rapid and specific LC-MS/MS method for the simultaneous quantification of T and TG in mouse plasma

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service