Skip to Content
Merck
All Photos(1)

Key Documents

SML2417

Sigma-Aldrich

ORG 25543 Hydrochloride

≥95% (HPLC)

Synonym(s):

4-Benzyloxy-3,5-dimethoxy-N-[(1-dimethylaminocyclopentyl)methyl]benzamide hydrochloride, N-[[1-(Dimethylamino)cyclopentyl]methyl]-3,5-dimethoxy-4-(phenylmethoxy)benzamide hydrochloride, ORG 25,543 hydrochloride, ORG 25543 hydrochloride, ORG-25543 hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H32N2O4·HCl
CAS Number:
Molecular Weight:
448.98
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥95% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

room temp

SMILES string

Cl.N(C3(CCCC3)CNC(=O)c1cc(c(c(c1)OC)OCc2ccccc2)OC)(C)C

InChI

1S/C24H32N2O4.ClH/c1-26(2)24(12-8-9-13-24)17-25-23(27)19-14-20(28-3)22(21(15-19)29-4)30-16-18-10-6-5-7-11-18;/h5-7,10-11,14-15H,8-9,12-13,16-17H2,1-4H3,(H,25,27);1H

InChI key

NIPQJILJYQVZJR-UHFFFAOYSA-N

Biochem/physiol Actions

Brain-penetrant, high-affinity, potent and selective glycine transporter 2 (GlyT-2; GlyT2) inhibitor with higher potency and selectivity than ALX-1393.
ORG 25543 is a brain-penetrant (free brain/plasma ratio = 0.53; 35 min post 2 or 20 mg/kg i.v. in mice), high-affinity, potent and selective glycine transporter 2 (GlyT-2; GlyT2) inhibitor (human & mouse pIC50 = 7.9/ GlyT2 vs <4/GlyT1) with great selectivity over a panel of 56 receptor and channel proteins. ORG 25543 is more potent and selective than the brain-impermeable ALX-1393 (GlyT2/GlyT1 pIC50 = 7.1/5.4) and exhibits high in vivo efficacy in a murine diabetic neuropathic pain model (ED50 = 0.07-0.16 mg/kg i.v.; Emin = 0.01 mg/kg). ORG 25543 is practically irreversible due to its tight-binding nature, suboptimal dosage should be applied in vivo to allow low target occupancy only and minimize acute toxicity.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Katsuya Morita et al.
The Journal of pharmacology and experimental therapeutics, 326(2), 633-645 (2008-05-02)
Neuropathic pain is refractory against conventional analgesics, and thus novel medicaments are desired for the treatment. Glycinergic neurons are localized in specific brain regions, including the spinal cord, where they play an important role in the regulation of pain signal
A Mingorance-Le Meur et al.
British journal of pharmacology, 170(5), 1053-1063 (2013-08-22)
Available medications for chronic pain provide only partial relief and often cause unacceptable side effects. There is therefore a need for novel molecular targets to develop new therapeutics with improved efficacy and tolerability. Despite encouraging efficacy data in rodents with
M A Gradwell et al.
The Journal of physiology, 595(23), 7185-7202 (2017-09-15)
Spinal parvalbumin-expressing interneurons have been identified as a critical source of inhibition to regulate sensory thresholds by gating mechanical inputs in the dorsal horn. This study assessed the inhibitory regulation of the parvalbumin-expressing interneurons, showing that synaptic and tonic glycinergic
Cristina Romei et al.
Neurochemistry international, 99, 169-177 (2016-07-11)
Glycine can be substrate for two transporters: GlyT1, largely expressed by astrocytes but also by some non-glycinergic neurons, and GlyT2, most frequently present in glycine-storing nerve endings. In morphological studies, GlyT2 expression had been found to be restricted to caudal
The first potent and selective inhibitors of the glycine transporter type 2.
W L Caulfield et al.
Journal of medicinal chemistry, 44(17), 2679-2682 (2001-08-10)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service