SML2220
SGC-AAK1-1N
≥98% (HPLC)
Synonym(s):
N-(6-(3-(Cyclopropanesulfonamido)phenyl)-1H-indazol-3-yl)isobutyramide, SGC-AAK1-1 Negative Control
About This Item
Recommended Products
Assay
≥98% (HPLC)
form
powder
color
white to beige
solubility
DMSO: 2 mg/mL, clear
storage temp.
2-8°C
SMILES string
O=C(C(C)C)NC1=NNC2=C1C=CC(C3=CC(NS(C4CC4)(=O)=O)=CC=C3)=C2
InChI
1S/C20H22N4O3S/c1-12(2)20(25)21-19-17-9-6-14(11-18(17)22-23-19)13-4-3-5-15(10-13)24-28(26,27)16-7-8-16/h3-6,9-12,16,24H,7-8H2,1-2H3,(H2,21,22,23,25)
InChI key
RAIAORGFMNXPOV-UHFFFAOYSA-N
Biochem/physiol Actions
SGC-AAK1-1, the active probe, is available from Sigma. To learn more about and purchase SGC-AAK1-1, click here.
To learn about other SGC chemical probes, visit sigma.com/sgc
Features and Benefits
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service