Skip to Content
Merck
All Photos(1)

Key Documents

SML1713

Sigma-Aldrich

Carprofen

≥97% (HPLC)

Synonym(s):

6-Chloro-α-methyl-9H-carbazole-2-acetic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H12ClNO2
CAS Number:
Molecular Weight:
273.71
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77
Pricing and availability is not currently available.

Quality Level

Assay

≥97% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 20 mg/mL, clear

storage temp.

room temp

SMILES string

CC(C(O)=O)c1ccc2c(c1)[nH]c3ccc(Cl)cc23

InChI

1S/C15H12ClNO2/c1-8(15(18)19)9-2-4-11-12-7-10(16)3-5-13(12)17-14(11)6-9/h2-8,17H,1H3,(H,18,19)

InChI key

PUXBGTOOZJQSKH-UHFFFAOYSA-N

Gene Information

human ... PTGS2(5743)

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Carprofen is a non-steroidal anti-inflammatory drug (NSAID) that has been found to have antimicrobial activity. Carprofen is primarily used as a veterinary analgesic and anti-inflammatory for arthritis and pain. Its anti-inflammatory activity is due to cyclooxygenase inihbition with selectivity for COX-2 inhibition, while its antimicrobial activity is less certain. Carprofen can kill B. subtilis by permeabilizing its membrane. Other studies have shown carprofen can target the Escherichia coli DNA polymerase III β subunit.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Arundhati Maitra et al.
British medical bulletin, 118(1), 138-148 (2016-05-07)
The number of cases of drug-resistant Mycobacterium tuberculosis, the causative agent of tuberculosis (TB), has risen rapidly in recent years. This has led to the resurgence in repurposing existing drugs, such as non-steroidal anti-inflammatory drugs (NSAIDs), for anti-TB treatment. Evidence
Zhou Yin et al.
Chemistry & biology, 21(4), 481-487 (2014-03-19)
Evidence suggests that some nonsteroidal anti-inflammatory drugs (NSAIDs) possess antibacterial properties with an unknown mechanism. We describe the in vitro antibacterial properties of the NSAIDs carprofen, bromfenac, and vedaprofen, and show that these NSAIDs inhibit the Escherichia coli DNA polymerase III
Anne Lamsa et al.
ACS chemical biology, 11(8), 2222-2231 (2016-05-20)
Increasing antimicrobial resistance has become a major public health crisis. New antimicrobials with novel mechanisms of action (MOA) are desperately needed. We previously developed a method, bacterial cytological profiling (BCP), which utilizes fluorescence microscopy to rapidly identify the MOA of
San Min Leow et al.
American journal of obstetrics & gynecology MFM, 2(2), 100084-100084 (2020-12-22)
Accurate prediction of spontaneous preterm labor/preterm birth in asymptomatic women remains an elusive clinical challenge because of the multi-etiological nature of preterm birth. The aim of this study was to develop and validate an immunoassay-based, multi-biomarker test to predict spontaneous preterm
Suad M Abdirahman et al.
Cancers, 12(9) (2020-08-23)
Colorectal cancer (CRC) is a challenging disease, with a high mortality rate and limited effective treatment options, particularly for late-stage disease. Patient-derived xenografts (PDXs) have emerged as an informative, renewable experimental resource to model CRC architecture and biology. Here, we

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service