Skip to Content
Merck
All Photos(1)

Key Documents

SAE0215

Sigma-Aldrich

Inorganic Pyrophosphatase from Escherichia coli

≥100 units/mL, buffered aqueous solution

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352204

biological source

Escherichia coli

Quality Level

recombinant

expressed in E. coli

grade

for molecular biology

description

Recombinant, expressed in E.coli

Assay

≥95% (size exclusion chromatography)

form

buffered aqueous solution

specific activity

≥100 units/mL

shelf life

2 yr at -20 °C ((retest))

mol wt

19.7 kDa

storage condition

OK to freeze

concentration

≥100 units/mL

color

colorless

optimum pH

9.0 (25 °C)

pH

8.0 (25 °C)

solubility

soluble
water: miscible

suitability

suitable for molecular biology

UniProt accession no.

application(s)

research use

foreign activity

DNAse, none detected
RNAse, none detected
Nickase, none detected

shipped in

dry ice

storage temp.

-10 to -25°C

General description

Inorganic pyrophosphatase (PPase) is a ubiquitous enzyme that catalyzes pyrophosphate hydrolysis. It plays an important role in energy metabolism by providing a thermodynamic pull for biosynthetic reactions, such as protein, RNA, and DNA synthesis. Nucleic acid synthesis would be energetically impossible in vivo if not coupled with the hydrolysis of pyrophosphate (PPi).

Application

This product is based on the native pyrophosphatase from E. coli, Uniprot No. P0A7A9. Pyrophosphatase in E. coli is a homohexameric protein containing 175 amino acids residues per subunit. This product is a recombinant protein expressed in E. coli and induced by IPTG. Each subunit has a MW of 19.7 kDa and theoretical pI of ~5. The protein activity is Mg2+ dependent and it is a relatively thermostable protein.

Biochem/physiol Actions

Inorganic pyrophosphatase (PPase) is a ubiquitous enzyme that catalyzes pyrophosphate to phosphate. It plays an important role in energy metabolism as it provides a thermodynamic pull for biosynthetic reactions, such as protein, RNA, and DNA synthesis.

Features and Benefits

This product has a purity minimum of 95% (SEC-HPLC) and an activity minimum of 100 units/mL to enhance RNA yield during transcription.

Unit Definition

One unit will release 1.0 µmole of inorganic orthophosphate per minute at pH 9 at 25 °C. The reaction buffer used for determination of enzyme activity contains 50 mM Tris-HCl, pH 9.0.

Physical form

The product is supplied as an aqueous solution containing 20mM Tris-HCl, 100mM NaCl, 1mM DTT, 0.1mM EDTA, and 50% glycerol, titrated to pH 8 at 25 °C.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Andrew C Pratt et al.
Journal of structural biology, 192(1), 76-87 (2015-08-25)
Family I inorganic pyrophosphatases (PPiases) are ubiquitous enzymes that are critical for phosphate metabolism in all domains of life. The detailed catalytic mechanism of these enzymes, including the identity of the general base, is not fully understood. We determined a
A Salminen et al.
The Journal of biological chemistry, 274(48), 33898-33904 (1999-11-24)
A homohexameric molecule of Escherichia coli pyrophosphatase is arranged as a dimer of trimers, with an active site present in each of its six monomers. Earlier we reported that substitution of His(136) and His(140) in the intertrimeric subunit interface splits
Kyung Min Ko et al.
FEBS letters, 581(28), 5445-5453 (2007-11-06)
Inorganic pyrophosphatase (PPase) catalyzes the hydrolysis of inorganic pyrophosphate (PPi) into phosphate (Pi), which provides a thermodynamic driving force for important biosynthetic reactions. The nematode Caenorhabditis elegans gene C47E12.4 encodes a PPase (PYP-1) which shows 54% amino acid identity with
Microbial inorganic pyrophosphatases.
R Lahti
Microbiological reviews, 47(2), 169-178 (1983-06-01)
R Lahti et al.
Journal of bacteriology, 170(12), 5901-5907 (1988-12-01)
Escherichia coli K-12 gene ppa encoding inorganic pyrophosphatase (PPase) was cloned and sequenced. The 5' end of the ppa mRNA was identified by primer extension mapping. A typical E. coli sigma 70 promoter was identified immediately upstream of the mRNA

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service