Skip to Content
Merck
All Photos(1)

Key Documents

SAB4503192

Sigma-Aldrich

Anti-SLC24A6, C-Terminal antibody produced in rabbit

affinity isolated antibody

Synonym(s):

NCKX6, Na(+)/K(+)/Ca(2+)-exchange protein 6, SLC24A6, Sodium/potassium/calcium exchanger 6, Solute carrier family 24 member 6

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 64 kDa

species reactivity

human

concentration

~1 mg/mL

technique(s)

ELISA: 1:20000
western blot: 1:500-1:1000

NCBI accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SLC24A6(80024)

General description

The SLC24A6 (solute carrier family 8 member B1) gene is mapped to human chromosome 12q24.13. The gene codes for a K+-dependent Na+/Ca2+ exchanger. The encoded protein is ubiquitously expressed in all the tissues. It is localized to inner mitochondrial membrane.

Immunogen

The antiserum was produced against synthesized peptide derived from human SLC24A6.

Immunogen Range: 494-543

Biochem/physiol Actions

SLC24A6 (solute carrier family 8 member B1) protein is responsible for maintaining Ca2+ homeostasis in the cell. SLC24A6 protein regulates sodium uptake and calcium efflux from the mitochondria.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Privileged crosstalk between TRPV1 channels and mitochondrial calcium shuttling machinery controls nociception.
Nita I I, et al.
Biochimica et Biophysica Acta, 1863(12), 2868-2880 (2016)
NCLX Protein, but Not LETM1, Mediates Mitochondrial Ca2+ Extrusion, Thereby Limiting Ca2+-induced NAD(P)H Production and Modulating Matrix Redox State*
De Marchi U, et al.
The Journal of Biological Chemistry, 289(29), 20377-20385 (2014)
Molecular cloning of a sixth member of the K+-dependent Na+/Ca2+ exchanger gene family, NCKX6.
Cai X, et al.
The Journal of Biological Chemistry, 279(7), 5867- 5876 (2004)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service