Skip to Content
Merck
All Photos(4)

Key Documents

SAB3500393

Sigma-Aldrich

Anti-Wnt10a antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-Wingless-type MMTV integration site family 10a

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

predicted mol wt 46 kDa

species reactivity

human, rat, mouse

technique(s)

immunofluorescence: suitable
immunohistochemistry: suitable
indirect ELISA: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... WNT10A(80326)

Immunogen

Wnt10a antibody was raised against a 14 amino acid peptide from near the carboxy terminus of human Wnt10a.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Linkage

The action of this antibody can be blocked using blocking peptide SBP3500393.

Physical form

Supplied in PBS with 0.02% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

related product

Product No.
Description
Pricing

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Naoya Matsunaga et al.
Cancer research, 78(13), 3698-3708 (2018-05-08)
Although malignant phenotypes of triple-negative breast cancer (TNBC) are subject to circadian alterations, the role of cancer stem cells (CSC) in defining this circadian change remains unclear. CSC are often characterized by high aldehyde dehydrogenase (ALDH) activity, which is associated
James W Foster et al.
Investigative ophthalmology & visual science, 62(6), 16-16 (2021-05-15)
To identify global gene expression changes in the corneal epithelium of keratoconus (KC) patients compared to non-KC myopic controls. RNA-sequencing was performed on corneal epithelium samples of five progressive KC and five myopic control patients. Selected results were validated using
Akihiro Kuma et al.
PloS one, 9(7), e103240-e103240 (2014-07-24)
WNT signaling mediates various physiological and pathological processes. We previously showed that WNT10A is a novel angio/stromagenic factor involved in such processes as tumor growth, wound healing and tissue fibrosis. In this study, we investigated the role of WNT10A in
Keishi Oda et al.
Respiratory research, 17, 39-39 (2016-04-14)
WNT/β-catenin signaling plays an important role in the pathogenesis of idiopathic pulmonary fibrosis (IPF); however, the role of WNT10A via transforming growth factor (TGF)-β signaling remains unclear. We evaluated the expression of WNT10A and TGF-β in bleomycin (BLM)-treated mice and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service