Skip to Content
Merck
All Photos(4)

Key Documents

SAB3500269

Sigma-Aldrich

Anti-Survivin antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-TIAP

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

predicted mol wt 16 kDa

species reactivity

human, mouse

technique(s)

immunocytochemistry: suitable
immunofluorescence: suitable
immunohistochemistry: suitable
indirect ELISA: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

Gene Information

human ... BIRC5(332)

Immunogen

Survivin antibody was raised against a peptide corresponding to amino acids near the amino terminus of human survivin.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Linkage

The action of this antibody can be blocked using blocking peptide SBP3500269.

Physical form

Supplied in PBS with 0.02% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

related product

Product No.
Description
Pricing

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fang Xu et al.
Oncology letters, 15(4), 5871-5878 (2018-03-20)
Multiple myeloma (MM) is a malignancy of the bone marrow. The median survival time of patients with MM is only 5 years, with patients frequently experiencing relapse. Currently, there is no effective therapy for recurrent MM. The results of the
X-L Cheng et al.
European review for medical and pharmacological sciences, 18(6), 769-774 (2014-04-08)
To explore the effect of edaravone (ED) on apoptosis of hippocampus neurons in seizures rats induced by pentylenetetrazole (PTZ). Forty-eight adult Wistar rats were randomly divided into normal control (NC) group, PTZ group, and ED group. A dose of PTZ
Siqi Wang et al.
Experimental and therapeutic medicine, 14(2), 1212-1218 (2017-08-16)
Colorectal cancer has a relatively low sensitivity to paclitaxel. The purpose of this study was to investigate the role of connexin 43 (Cx43), which is a structural component of gap junctional communication (GJC), in paclitaxel cytotoxicity in colorectal cancer cells.
Amr A Fouad et al.
Environmental toxicology and pharmacology, 38(2), 420-429 (2014-08-17)
The protective effect of naringenin, a flavonoid compound isolated from citrus fruits, was investigated against nephrotoxicity induced by gentamicin (80mgkg(-1)/day, i.p., for eight days) in rats. Naringenin treatment (50mgkg(-1)/day, p.o.) was administered for eight days, starting on the same day
Wenyan Lu et al.
Journal of cellular biochemistry, 115(10), 1799-1807 (2014-06-07)
Emerging evidence indicates that activation of Wnt/β-catenin signaling at the cell surface results in inhibition of glycogen synthase kinase 3β (GSK3β), leading to activation of mTORC1 signaling in cancer cells. The low density lipoprotein receptor-related protein-6 (LRP6) is an essential

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service